Skip to Content
Merck
All Photos(1)

Key Documents

51456

Sigma-Aldrich

Urea

BioUltra, for molecular biology, 99% (T)

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

99% (T)

form

powder or crystals

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.1% biuret
<0.005% formaldehyde (HCHO)

ign. residue

≤0.01% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

pH

8.0-10.0 (20 °C, 8 M in H2O)

mp

132-135 °C (lit.)

solubility

H2O: 8 M at 20 °C, clear, colorless

density

1.335 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NC(N)=O

λ

8 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.02

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Other Notes

Denaturing reagent; Commonly used in conjunction with polyacrylamide gel electrophoresis for the fractionation of single-stranded nucleic acids with chains of up to 200 nucleotides

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fractionation of low molecular weight DNA or RNA in polyacrylamide gels containing 98% formamide or 7 M urea.
T Maniatis et al.
Methods in enzymology, 65(1), 299-305 (1980-01-01)
R. Poulson et al.
The Ribonucleic Acids, 333-333 (1977)
A Paulus et al.
Electrophoresis, 14(1-2), 27-35 (1993-01-01)
DNA digest analysis with polymer-filled capillaries in capillary electrophoresis is described. The samples analyzed consisted of commercially available standards including a 100 base pair ladder with repeating units up to 2000 base pairs. Three DNA digests covered the most common
R.C. Ogden et al.
Methods in Enzymology, 152, 66-66 (1987)
Gurudayal et al.
Nano letters, 15(6), 3833-3839 (2015-05-06)
Photoelectrochemical water splitting half reactions on semiconducting photoelectrodes have received much attention but efficient overall water splitting driven by a single photoelectrode has remained elusive due to stringent electronic and thermodynamic property requirements. Utilizing a tandem configuration wherein the total

Protocols

An optimized LC-MS/MS based workflow for low artifact tryptic digestion and peptide mapping of monoclonal antibody, adalimumab (Humira) using filter assisted sample preparation (FASP).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service