Skip to Content
Merck
All Photos(1)

Key Documents

1672206

USP

Topiramate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,3:4,5-Bis-O-(1-methylethylidene)-36-D-fructo-pyranose sulfamate, McN 4853, RWJ 17021, Topamax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO8S
CAS Number:
Molecular Weight:
339.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

topiramate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NS(OC[C@]12[C@](OC(C)(C)O2)([H])[C@@]3([H])[C@@](OC(C)(C)O3)([H])CO1)(=O)=O

InChI

1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1

InChI key

KJADKKWYZYXHBB-XBWDGYHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Topiramate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Topiramate Capsules
  • Topiramate Tablets
  • Topiramate Compounded Oral Suspension
Topiramate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Topiramate Tablets
  • Topiramate Compounded Oral Suspension

Biochem/physiol Actions

Kainate GluR5 receptor antagonist; anticonvulsant.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topiramate Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 34(5), 4434-4434 (2020)
Topiramate Compounded Oral Suspension
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(4), 4436-4436 (2014)
Topiramate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 4429-4429 (2020)
Topiramate Capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(1), 4432-4432 (2020)
Mattias Linde et al.
The Cochrane database of systematic reviews, 6(6), CD010610-CD010610 (2013-06-26)
Some antiepileptic drugs but not others are useful in clinical practice for the prophylaxis of migraine. This might be explained by the variety of actions of these drugs in the central nervous system. The present review is part of an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service