Skip to Content
Merck
All Photos(1)

Key Documents

1603006

USP

Riboflavin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

riboflavin

manufacturer/tradename

USP

mp

290 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
R0600000R7649Y0000757
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

API family

riboflavin

API family

riboflavin

API family

-

API family

riboflavin

mp

290 °C (dec.) (lit.)

mp

290 °C (dec.) (lit.)

mp

290 °C (dec.) (lit.)

mp

290 °C (dec.) (lit.)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Riboflavin USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, and standard stock Riboflavin solution for assay, identification, and performance test according to the given below monographs and general chapters of United States Pharmacopeia (USP):
  • Riboflavin[1]
  • Riboflavin Tablets[2]
  • Water-Soluble Vitamins Preparation[3]
  • 〈481〉 Riboflavin Assay[4]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water-Soluble Vitamins Preparation
United States Pharmacopeia, 45(5) (2021)
Riboflavin Tablets
United States Pharmacopeia, 41(6), 3870-3870 (2017)
?481? Riboflavin Assay
United States Pharmacopeia, 41(6), 6735-6735 (2017)
Riboflavin
United States Pharmacopeia, 30(3), 3869-3869 (2013)
Christine Wittig-Silva et al.
Ophthalmology, 121(4), 812-821 (2014-01-08)
To report the refractive, topographic, and clinical outcomes 3 years after corneal collagen cross-linking (CXL) in eyes with progressive keratoconus. Prospective, randomized controlled trial. One hundred eyes with progressive keratoconus were randomized into the CXL treatment or control groups. Cross-linking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service