Skip to Content
Merck
All Photos(1)

Key Documents

47861

Supelco

Riboflavin (B2)

analytical standard

Synonym(s):

(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E101

grade

analytical standard

Quality Level

form

solid

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

290 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Riboflavin (B2) is a vitamin, which is a vital component of mitochondrial energy production. It can be used in the treatment of mitochondrial dysfunction and also for the prophylactic treatment of migraine in adults. It is a part of the coenzyme flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD), involved in a number of oxidation and reduction reactions.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Riboflavin (B2) may be used as an analytical reference standard for the quantification of the analyte in neutraceutical products and food products using high-performance liquid chromatography technique.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norman.AR
Norman.AR
Diagnosis of Aging Skin Diseases (2008)
The use of nutraceutics in children`s and adolescent?s headache
Sangermani.R and Boncimino.A
Neurological Sciences, 38(1), 121-124 (2017)
Determination of seven water-soluble vitamins in tarhana, a traditional Turkish cereal food, by high-performance liquid chromatography
Ekinci R and Kadakal C
Acta Chromatographica , 15, 289-289 (2005)
Quantitative simultaneous estimation of water-soluble vitamins, Riboflavin, Pyridoxine, Cyanocobalamin and Folic Acid in neutraceutical products by HPLC
Perveen S, et al.
The Open Analytical Chemistry Journal, 3, 1-5 (2009)
Carol P Wilson et al.
Hypertension (Dallas, Tex. : 1979), 61(6), 1302-1308 (2013-04-24)
Intervention with riboflavin was recently shown to produce genotype-specific lowering of blood pressure (BP) in patients with premature cardiovascular disease homozygous for the 677C→T polymorphism (TT genotype) in the gene encoding the enzyme methylenetetrahydrofolate reductase (MTHFR). Whether this effect is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service