Skip to Content
Merck
All Photos(1)

Key Documents

Y0000757

Riboflavin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

riboflavin

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

290 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Riboflavin, also known as vitamin B2, is crucial for human nutrition, and growth. It finds potential applications in biological redox reactions and is found in several foodstuffs, energy drinks, and vitamin formulations.[1][2]

Application

Riboflavin may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.[3]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riboflavin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 5(2), 3484-3485 (2008)
Differential pulse polarographic method development and validation of riboflavin in pharmaceutical formulation
Kashid LM and Patil SV
International Journal of Scientific Research, 5(2), 325-334 (2015)
Simultaneous determination of riboflavin phosphate and other ingredients in a multivitamin pharmaceutical preparation by on-line automated LC coupled with pre-column immobilized enzyme reactor
Ono M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(1-2), 325-334 (2002)
Olivier Richoz et al.
Investigative ophthalmology & visual science, 55(9), 5783-5787 (2014-07-24)
When treating peripheral ectatic disease-like pellucid marginal degeneration (PMD), corneal cross-linking with UV-A and riboflavin (CXL) must be applied eccentrically to the periphery of the lower cornea, partly irradiating the corneal limbus. Here, we investigated the effect of standard and
Charles A Abbas et al.
Microbiology and molecular biology reviews : MMBR, 75(2), 321-360 (2011-06-08)
Riboflavin [7,8-dimethyl-10-(1'-d-ribityl)isoalloxazine, vitamin B₂] is an obligatory component of human and animal diets, as it serves as the precursor of flavin coenzymes, flavin mononucleotide, and flavin adenine dinucleotide, which are involved in oxidative metabolism and other processes. Commercially produced riboflavin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service