Skip to Content
Merck
All Photos(2)

Key Documents

57291-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 7 μm (PDMS)

Synonym(s):

PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

product name

SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler

material

fused silica fiber
plain green hub

Quality Level

needle size

23 ga

packaging

pkg of 3 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

7 μm (PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS coating

application(s)

food and beverages

compatibility

for analyte group volatiles (MW 80-500)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Features and Benefits

Designed for use with Merlin Microseal sealing system; can be used with other septumless systems.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu et al.
Journal of chromatographic science, 38(9), 377-382 (2000-09-30)
An empirical model describing the relationship between the partition coefficients (K) of perfume materials in the solid-phase microextraction (SPME) fiber stationary phase and the Linearly Temperature Programmed Retention Index (LTPRI) is obtained. This is established using a mixture of eleven
Automatic Injection SPME-Chiral-GC-MSD Analysis of Essential Oils
Coleman, W., et al.
Journal of Chromatographic Science, 36, 575-578 (1998)
Characterization of Roasted Coffee and Coffee Beverages by SPME-GC and Principal Component Analysis
Bicchi, C., et al.
Journal of Agricultural and Food Chemistry, 45, 4680-4686 (1997)
Interaction Between Natural Organic Matter (NOM) and (PAH) Comparison of Fluorescence Quenching and SPME
Doll TE, et al.
Fresenius Journal of Analytical Chemistry, 364(4), 313-319 (1999)
M F Sledge et al.
Journal of chromatography. A, 873(1), 73-77 (2000-04-11)
Solid-phase microextraction has been used to investigate chemical communication in several social wasp species. Using the technique to analyse exocrine gland secretions, we demonstrate that the results are comparable with those obtained with the more classical methods that use solvents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service