Skip to Content
Merck
All Photos(1)

Key Documents

PHR1518

Supelco

Polydimethylsiloxane

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dimethylpolysiloxane, Polydimethylsiloxane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. S0600000
traceable to USP 1546300

API family

polydimethylsiloxane

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.

Application

It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7823 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robust and biodegradable elastomers based on corn starch and polydimethylsiloxane (PDMS).
Ceseracciu L
ACS Applied Materials & Interfaces, 7(6), 3742-3753 (2015)
Molecular weight distribution of poly (dimethylsiloxane) by combining matrix-assisted laser desorption/ionization time-of-flight mass spectrometry with gel-permeation chromatography fractionation.
Montaudo, Giorgio, et al.
Rapid Communications in Mass Spectrometry, 9.12, 1158-1163 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service