Skip to Content
Merck
All Photos(2)

Key Documents

57289-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 30 μm (PDMS)

Synonym(s):

PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

product name

SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 23 ga, for use with autosampler

material

fused silica fiber
plain yellow hub

Quality Level

needle size

23 ga

packaging

pkg of 3 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS coating

application(s)

food and beverages

compatibility

for analyte group volatiles (MW 80-500)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Features and Benefits

Designed for use with Merlin Microseal sealing system; can be used with other septumless systems.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-Phase Microextraction for Cannabinoids Analysis in Hair and Its Possible Application to Other Drugs
Strano-Rossi, S., et al.
Journal of Analytical Toxicology, 23 (1999)
B J Hall et al.
Analytical chemistry, 70(9), 1788-1796 (1998-05-26)
Solid-phase microextraction (SPME) is applied to the determination of cannabidiol, delta 8-tetrahydrocannabinol (delta 8-THC), delta 9-tetrahydrocannabinol (delta 9-THC), and cannabinol in pure water and human saliva. The inherent extraction behavior of the cannabinoids in pure water is evaluated along with
L van der Wal et al.
Chemosphere, 54(4), 561-568 (2003-10-29)
Uptake and accumulation of three chlorobenzenes was studied in both biota (Enchytraeus crypticus) and 30 mum polydimethylsiloxane (PDMS) solid phase microextraction (SPME) fibers after exposure to spiked OECD soil. The OECD soil was spiked with three different concentrations of all
Bruce A Tomkins et al.
Journal of chromatography. A, 964(1-2), 21-33 (2002-08-30)
A rapid, sensitive, convenient, and highly quality-assured method is presented for the determination of 19 organochlorine pesticides (OCPs) in small samples (10 ml) of ground water. Samples are initially fortified with 2,4,5,6-tetrachloro-m-xylene (surrogate) and decachlorobiphenyl (retention time marker), then extracted
Online Determination of Organophosphorus Pesticides in Water by SPME and GC with Thermionic-Selective Detection
Lopez-Avial, V., et al.
Journal of High Resolution Chromatography, 20 (9), 487-492 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service