57184
Polyethylene (PE) Frit, 20 μm porosity
for use with 60mL SPE Tube, pk of 32
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
description
for SPE Tube
Quality Level
packaging
pk of 32
technique(s)
solid phase extraction (SPE): suitable
pore size
20 μm
application(s)
food and beverages
compatibility
for use with 60mL SPE Tube
Looking for similar products? Visit Product Comparison Guide
General description
Frits are crucial components of the high performance liquid chromatography system design and act as a depth filter for the concurrent physical removal of small molecules. Frits are available in various materials. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service