57181
Polyethylene (PE) Frit, 20 μm porosity
for use with 6mL SPE Tube, pk of 60
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
material
polyethylene tube
Quality Level
description
for SPE Tube
packaging
pk of 60
technique(s)
solid phase extraction (SPE): suitable
pore size
20 μm
application(s)
food and beverages
compatibility
for use with 6mL SPE Tube
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service