Skip to Content
Merck
All Photos(1)

Key Documents

57098

Supelco

Female Luer Cap

polypropylene, configured for capping luer tips, pkg of 12 ea

Synonym(s):

Luer Plugs & Caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

polypropylene

Quality Level

packaging

pkg of 12 ea

technique(s)

solid phase extraction (SPE): suitable

application(s)

environmental
food and beverages

compatibility

configured for capping luer tips
for use with Supelco Visidry Drying Attachment

Looking for similar products? Visit Product Comparison Guide

General description

Use to close Luer port. Polypropylene.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service