Key Documents
27473
Supelco
Crimp seals, 11 mm, with PTFE/rubber septa
red aluminum seal, seal diam. 11 mm, PTFE/red rubber septum, pkg of 1000 ea
About This Item
UNSPSC Code:
41121821
Pricing and availability is not currently available.
material
PTFE/red rubber septum
red aluminum seal
feature
closure type crimp seal
packaging
pkg of 1000 ea
seal diam.
11 mm
fitting
thread for 11 mm
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
85414027102-U854980-U
feature
closure type crimp seal
feature
closure type crimp seal
feature
closure type crimp seal
feature
closure type crimp seal
material
PTFE/red rubber septum, red aluminum seal
material
PTFE-faced rubber septum, silver aluminum seal
material
PTFE/red rubber septum, silver aluminum seal
material
septum (TFE/butyl/red rubber), silver aluminum seal
packaging
pkg of 1000 ea
packaging
pkg of 100 ea
packaging
pkg of 1 × 100 ea (27102-U)
packaging
pkg of × 1000 ea
seal diam.
11 mm
seal diam.
11 mm
seal diam.
-
seal diam.
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)