Skip to Content
Merck
All Photos(2)

Key Documents

M66005

Sigma-Aldrich

3-Methylpentane

≥99%

Synonym(s):

3-Methylpentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2CHCH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730734
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.97 (vs air)

Quality Level

vapor pressure

135 mmHg ( 17 °C)

Assay

≥99%

form

liquid

autoignition temp.

532 °F

expl. lim.

~7.7 %

refractive index

n20/D 1.376 (lit.)

bp

64 °C (lit.)

density

0.664 g/mL at 25 °C (lit.)

SMILES string

CCC(C)CC

InChI

1S/C6H14/c1-4-6(3)5-2/h6H,4-5H2,1-3H3

InChI key

PFEOZHBOMNWTJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methylpentane (3MP) is mainly used as a model n-alkane in studies relating to densities, viscosities, refractive indices, excess enthalpies and vapor-liquid equilibrium of binary alkane mixtures.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapor-liquid equilibriums for the binary systems n-hexane with 2-methylpentane, 3-methylpentane, and 2, 4-dimethylpentane.
Ho C L and Davison R R
Journal of Chemical and Engineering Data, 24(4), 293-296 (1979)
Densities, viscosities, and refractive indices of the binary systems methyl tert-butyl ether+ 2-methylpentane,+ 3-methylpentane,+ 2, 3-dimethylpentane, and+ 2, 2, 4-trimethylpentane at 298.15 K.
Bouzas A, et al.
Journal of Chemical and Engineering Data, 45(2), 331-333 (2000)
Richard C Bell et al.
The Journal of chemical physics, 127(2), 024704-024704 (2007-07-21)
An ion soft-landing approach was applied to study the motion of hydronium (D(3)O(+)) and cesium (Cs(+)) ions from 84 to 104 K in glassy 3-methyl-pentane (3MP) films vapor deposited on Pt(111). Both ions were found to have very similar mobilities
3-Methylpentane. CAS# 96-14-0.
J B Galvin et al.
Journal of toxicology and environmental health. Part A, 58(1-2), 93-102 (1999-10-28)
Shervin Shahriari et al.
The Journal of chemical physics, 121(18), 8960-8967 (2004-11-06)
We have measured the dielectric relaxation of several glass forming branched alkanes with very low dielectric loss in the frequency range 50 Hz-20 kHz. The molecular liquids of this study are 3-methylpentane, 3-methylheptane, 4-methylheptane, 2,3-dimethylpentane, and 2,4,6-trimethylheptane. All liquids display

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service