Skip to Content
Merck
All Photos(1)

Key Documents

M65807

Sigma-Aldrich

2-Methylpentane

≥99%

Synonym(s):

‘Isohexane’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Quality Level

vapor pressure

6.77 psi ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

583 °F

expl. lim.

7 %

refractive index

n20/D 1.371 (lit.)

bp

62 °C (lit.)

mp

−154 °C (lit.)

density

0.653 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylpentane is mainly used in studies involving the functionalization of aliphatic C–H bonds using different carbene insertion processes to form C–H insertion products. The metal-free Ritter-type amination reaction of tertiary C–H bond using iodic acid as an oxidant in the presence of N-hydroxyphthalimide has also been reported.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-Free C--H Functionalization of Alkanes by Aryldiazoacetates.
Tortoreto C, et al.
Organic Letters, 19(4), 770-773 (2017)
Ritter-type amination of C--H bonds at tertiary carbon centers using iodic acid as an oxidant.
Kiyokawa K, et al.
Chemical Communications (Cambridge, England), 52(89), 13082-13085 (2016)
Zinc (II)-Mediated carbene insertion into C--H bonds in alkanes.
Kulkarni N V, et al.
Inorganic Chemistry, 54(23), 11043-11045 (2015)
Andrea N Giordano et al.
The journal of physical chemistry. A, 119(15), 3545-3555 (2015-03-27)
We report the solvent-dependent dynamics of carbonyl site exchange for Fe(CO)3(η(4)-norbornadiene) (FeNBD) in a series of linear and nonlinear alkanes. The barrier to exchange is very low (∼1.5 kcal/mol), and the resulting carbonyl dynamics are rapid enough to lead to
2-Methylpentane (isohexane). CAS# 107-83-5.
J B Galvin et al.
Journal of toxicology and environmental health. Part A, 58(1-2), 81-92 (1999-10-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service