Z742700
BenchMixer™ V2 Vortexer with reversible top
AC/DC input 115 V AC, US 2-pin plug
Synonym(s):
vortex mixer, vortexer lab, vortexer mixer
All Photos(1)
About This Item
UNSPSC Code:
41103800
NACRES:
NB.73
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BV1003
Related Categories
General description
The BenchMixer V2 utilizes a smooth and powerful mixing technology similar to the original BenchMixer, with the advantage of a uniquely designed reversible rubber head that can be attached to the mixer in two orientations.
For the vortexing of single tubes, the cupped surface would generally be preferred. However, for applications requiring a flat/dimpled surface for mixing uniquely shaped vessels (or a handful of multiple test-tubes), the head can easily be flipped over and reattached in reverse orientation.
Like the original BenchMixer, the drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. With a maximum speed up to 3,200 rpm, the mixer provides an instant vortex of even the largest sample sizes, including full 50ml tubes.
For the vortexing of single tubes, the cupped surface would generally be preferred. However, for applications requiring a flat/dimpled surface for mixing uniquely shaped vessels (or a handful of multiple test-tubes), the head can easily be flipped over and reattached in reverse orientation.
Like the original BenchMixer, the drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. With a maximum speed up to 3,200 rpm, the mixer provides an instant vortex of even the largest sample sizes, including full 50ml tubes.
Features and Benefits
Reversible top platform, cup or flat-dimpled surface
Powerful motor for INSTANT vortexing
Continuous or "touch" operation
Q-Drive dynamic balancing system
Variable speed from 200 to 3200 rpm
Wide variety of accessories, for tubes up to 50ml
Powerful motor for INSTANT vortexing
Continuous or "touch" operation
Q-Drive dynamic balancing system
Variable speed from 200 to 3200 rpm
Wide variety of accessories, for tubes up to 50ml
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)