Key Documents
Z258423
Sigma-Aldrich
Vortex-Genie® 2 mixer
AC/DC input 220 V AC, Schuko plug
Synonym(s):
Laboratory vortex mixer, Vortex mixer
About This Item
UNSPSC Code:
41103800
NACRES:
NB.73
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Scientific Industries SI-0256
Scientific Industries
General description
Mixes liquids quickly and thoroughly with vortex action. Variable speed control allows precise mixing range. The unit contains a standard 3 in. platform head for flasks and beakers, and a cup head for single tube mixing. Accessories for special applications are sold separately. Speed: 600 to 3,200 rpm. Max. load: 1 lb.
  • Variable speed control
  • Shaking or vortex action
  • Continuous or touch-control mixing
Accessories sold separately
Legal Information
Vortex-Genie is a registered trademark of Scientific Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)