Skip to Content
Merck
All Photos(1)

Key Documents

SRP3109

Sigma-Aldrich

MCP-1/MCAF human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCL2, MCAF, Monocyte Chemotactic Protein-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL2(6347)

General description

Monocyte chemoattractant protein-1/monocyte chemotactic and activating factor (MCP-1/MCAF), also called the C-C motif chemokine ligand 2 (CCL2), is an inflammatory cytokine. The CCL2 gene is mapped to human chromosome 17q12. CCL2 is present in epithelial, endothelial, mesangial and microglial cells. Structurally, it dimerizes and has four regions of the ß-sheet. Recombinant human MCP-1 is a 8.6 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues. The MCP family cross-reacts across species.

Application

Monocyte chemoattractant protein-1/monocyte chemotactic and activating factor (MCP-1/MCAF) has been used:
  • as a chemokine supplement in the alpha minimal essential medium for culturing neurons to test its effect on JmjC histone demethylase (Jmjd2a) mRNA
  • as a component of Dulbecco′s Modified Eagle (DMEM) medium, as a positive chemoattractant control in HeLa cells for migration assay
  • as a chemoattractant to induced macrophage chemotaxis

Biochem/physiol Actions

Monocyte chemoattractant protein-1/monocyte chemotactic and activating factor (MCP-1/MCAF) acts as a human immunodeficiency virus 1 (HIV-1) suppressor. The MCP-1 acts as a chemoattractant for T cells and natural killer (NK) cells. It also plays a key role in the migration and infiltration of macrophages. CCL2 shows under expression in high-grade ovarian serous carcinomas (HGOSCs) and is expressed less in epithelial ovarian cancer (EOC).
Monocyte chemoattractant protein-1/monocyte chemotactic and activating factor (MCP-1/MCAF) is a member of CC chemokine family. It triggers the activation of monocytes by triggering degranulation as well as respiratory burst activity in monocytes. It facilitates monocytes in inhibiting or enhancing the growth of specific brain tumor cell lines. It is found to be overexpressed in cases of atherosclerosis, idiopathic pulmonary fibrosis, rheumatoid arthritis, and delayed hypersensitivity reactions. It is also implicated in Kawasaki disease.

Sequence

QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Wojnarowicz et al.
Oncogenesis, 1, e27-e27 (2012-01-01)
The frequent loss of heterozygosity of chromosome (Chr) 17 in epithelial ovarian cancer (EOC), particularly high-grade ovarian serous carcinomas (HGOSCs), has been attributed to the disruption of known tumour suppressor genes, such as TP53 (17p13), as well as other genes
Expression of monocyte chemoattractant protein-1 in Kawasaki disease: the anti-inflammatory effect of gamma globulin therapy.
Asano T and Ogawa S
Scandinavian Journal of Immunology, 51, 98-103 (2000)
Satish L Deshmane et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 29(6), 313-326 (2009-05-16)
Chemokines constitute a family of chemoattractant cytokines and are subdivided into four families on the basis of the number and spacing of the conserved cysteine residues in the N-terminus of the protein. Chemokines play a major role in selectively recruiting
Yang Xiang et al.
Clinical epigenetics, 14(1), 32-32 (2022-03-02)
Recent studies have suggested that folic acid can restore abnormal DNA methylation and monocyte subset shifts caused by hyperhomocysteinemia (HHcy) and hyperlipidemia (HL). However, the exact mechanism of action is still not fully understood. In this study, we further investigated
A W S de Souza et al.
Clinical and experimental immunology, 178(2), 270-278 (2014-07-24)
The objective of this study is to evaluate urinary high mobility group box 1 (HMGB1) levels as markers for active nephritis in patients with anti-neutrophil cytoplasmic antibody (ANCA)-associated vasculitis (AAV) in comparison with urinary CD4(+) effector memory T cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service