MMA205
Millicell® µ-Migration Assay Kit
This Kit overcomes the limitations of traditional multiwell migration assays. Its microfluidic, low-volume technology promotes a stable, diffusion-generated concentration gradient that is consistently linear & lasts for more than 48 hours.
Synonym(s):
microfluidic cellular migration assay kit
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NB.13
Recommended Products
species reactivity
rat, mouse, human
manufacturer/tradename
Millicell®
technique(s)
cell based assay: suitable
single cell analysis: suitable
shipped in
dry ice
General description
- Cell migration occurs in a variety of biological processes includingwound healing, embryonic development, and immune response. Because of the importance of cell migration in angiogenesis, oncology, neurology, andespecially immunology, there is an increased interest in understanding theunderlying biological mechanisms at a single cell level.
- Traditional multiwell (Boyden chamber) migration assays can have limitations, such as: (1) gradients are not linear, well-established, or controlled, (2) quantificationof migratory cells is based on an endpoint measurement rather than a dynamicdetermination, and (3) imaging of the cells while they are migrating iscurrently not possible.
- The Millicell µ-Migration Assay Kit overcomes these limitations.
- Its microfluidic, low-volume technology promotes a stable, diffusion-generatedconcentration gradient that is consistently linear and lasts for more than 48 hours. Designed for video microscopy assays, the µ-Migration Slide is made froma plastic with high optical qualities similar to those of glass. At specifictime intervals, images of the observation area can be acquired, allowingreal-time monitoring and quantitative measurements of cell migration.
Features and Benefits
- Live cell tracking of single cell or cell population migration
- Establishes a stable and linear concentration gradient that lasts ≥ 48 hours
- Helps distinguish chemotaxis from random movement
- Allows for multiparametric analysis, including cell directionality and velocity
- Enhanced optical imaging of slow- and fast-migrating cells
- Analyze 3 chemoattractants in parallel for increased throughput and flexibility
- No need to spend time or money on optimization
- Ready to use, no assembly needed
Components
- Millicell® µ-Migration Slides: (Part No. CS204500) Four uncoated sterilized slides. Each slide contains 3 chambers.
- Millicell® µ-Migration Caps and Plugs: (Part No. CS204501) Forty-eight of each. Twelve caps and twelve plugs for each slide.
- Human Collagen IV: (Part No. CS204498) One 100 µg vial of 1 mg/mL Human Collagen IV.
- Acetic Acid Buffer: (Part No. CS204485) One vial containing 1.5 mL of 0.5 M acetic acid.
- BSA Solution: (Part No. CS204507) One vial containing 100 µL of 30% BSA solution.
- Beveled pipette tips: Fisherbrand SureOne 200 µL tips, Fisher Cat. No. 02-707-419. Two 96-tip boxes.
Storage and Stability
1. Millicell® µ-Migration Slides (Part No. CS204500). Store at room temperature.
2. Millicell µ-Migration Caps and Plugs (Part No. CS204501) Store at room temperature.
3. Human Collagen IV (Part No. CS204498) Store at -20ºC.
4. Acetic Acid Buffer (Part No. CS204485) Store at room temperature.
5. BSA Solution (Part No. CS204507) Store at -20ºC.
6. Beveled Pipette Tips. Store at room temperature.
2. Millicell µ-Migration Caps and Plugs (Part No. CS204501) Store at room temperature.
3. Human Collagen IV (Part No. CS204498) Store at -20ºC.
4. Acetic Acid Buffer (Part No. CS204485) Store at room temperature.
5. BSA Solution (Part No. CS204507) Store at -20ºC.
6. Beveled Pipette Tips. Store at room temperature.
Legal Information
Accutase is a registered trademark of Innovative Cell Technologies, Inc.
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Qiao Liao et al.
Acta pharmacologica Sinica, 40(10), 1314-1321 (2019-05-08)
Irisin, a myokine, is cleaved from the extracellular portion of fibronectin domain-containing 5 protein in skeletal muscle and myocardium and secreted into circulation as a hormone during exercise. Irisin has been found to exert protective effects against lung and heart injuries.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service