Skip to Content
Merck
All Photos(1)

Key Documents

P5333

Sigma-Aldrich

Pleiotrophin human

≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

HARP, HB-GAM, Heparin affinity regulatory peptide, Heparin-binding growth factor, Heparin-binding growth-associated molecule, PTN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

predicted mol wt 15.3 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PTN(5764)

General description

Pleiotrophin (PTN) is a multifunctional growth factor. The protein coded by the PTN gene is a member of the heparin-binding cytokines family. This gene is located on human chromosome 7. Pleiotrophin is also known as heparin-binding brain mitogen (HBBM), heparin-binding growth factor-8 (HBGF-8), heparin-binding growth-associated molecule (HB-GAM) and osteoblast-specific factor (OSF-1). This gene is highly expressed in brain, uterus, gut, muscle, lung and skin. Pleiotrophin mRNA is expressed in osteoblasts, chondrocytes, tumor cells, fibroblasts, astrocytes and Schwann cells.

Application

PTN is active in growth and development, and has mitogenic and neurite outgrowth activity. The gene for pleiotrophin is highly expressed in brain, uterus, gut, muscle, lung, and skin tissues

Biochem/physiol Actions

Pleiotrophin participates in growth and development. Pleiotrophin is mitogenic and displays neurite outgrowth activity. It stimulates tumor angiogenesis. There is extraordinary conservation between the amino acid sequences of bovine, human, and rat pleiotrophin.
Pleiotrophin may be the first member of a family of developmentally regulated cytokines. It is active in growth and development and the pleiotrophin gene is highly expressed in brain, uterus, gut, muscle, lung, and skin. Pleiotrophin mRNA is expressed in osteoblasts, chondrocytes, fibroblasts, astrocytes, Schwann cells, and tumor cells. Pleiotrophin is mitogenic and displays neurite outgrowth activity. It stimulates tumor angiogenesis. Pleiotrophin is also known as heparin-binding brain mitogen (HBBM), heparin-binding growth factor-8 (HBGF-8), heparin-binding growth-associated molecule (HB-GAM) and osteoblast-specific factor (OSF-1). There is extraordinary conservation between the amino acid sequences of bovine, human, and rat pleiotrophin.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The activity is measured in culture by the ability to enhance neurite outgrowth of cerebral cortical neurons from rat embryos.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pleiotrophin expression during odontogenesis.
Erlandsen H, et al.
The Journal of Histochemistry and Cytochemistry, 60(5), 366-375 (2012)
Pleiotrophin as a central nervous system neuromodulator, evidences from the hippocampus.
Gonzalez-Castillo C, et al.
Frontiers in Cellular Neuroscience, 8, 443-443 (2015)
Characterization of the human pleiotrophin gene. Promoter region and chromosomal localization.
Li Y S, et al.
The Journal of Biological Chemistry, 267(36), 26011-26016 (1992)
H Muramatsu et al.
Biochemical and biophysical research communications, 177(2), 652-658 (1991-06-14)
Midkine (MK) is the product of a retinoic acid responsive gene, MK, and is the initial member of a new family of heparin binding factors. Recombinant MK produced by an L-cell clone transfected with an MK cDNA was purified to
Heather A Himburg et al.
Nature medicine, 16(4), 475-482 (2010-03-23)
Hematopoietic stem cell (HSC) self-renewal is regulated by both intrinsic and extrinsic signals. Although some of the pathways that regulate HSC self-renewal have been uncovered, it remains largely unknown whether these pathways can be triggered by deliverable growth factors to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service