Skip to Content
Merck
All Photos(1)

Key Documents

SML0132

Sigma-Aldrich

Lanreotide acetate

≥98% (HPLC)

Synonym(s):

3-(2-Naphthalenyl)-D-alanyl-L-cysteinyl-L-tyrosyl-D-tryptophyl-L-lysyl-L-valyl-L-cysteinyl-L-threoninamide cyclic (2→7)-disulfide acetate, Angiopeptin acetate, BIM-23014C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H69N11O10S2 · xC2H4O2
CAS Number:
Molecular Weight:
1096.32 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)[C@@H]1NC(=O)[C@H](CCCCN)NC(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CSSC[C@H](NC1=O)C(=O)N[C@@H]([C@@H](C)O)C(N)=O)NC(=O)[C@H](N)Cc5ccc6ccccc6c5

InChI

1S/C54H69N11O10S2.C2H4O2/c1-29(2)45-54(75)63-44(53(74)65-46(30(3)66)47(57)68)28-77-76-27-43(62-48(69)38(56)23-32-15-18-33-10-4-5-11-34(33)22-32)52(73)60-41(24-31-16-19-36(67)20-17-31)50(71)61-42(25-35-26-58-39-13-7-6-12-37(35)39)51(72)59-40(49(70)64-45)14-8-9-21-55;1-2(3)4/h4-7,10-13,15-20,22,26,29-30,38,40-46,58,66-67H,8-9,14,21,23-25,27-28,55-56H2,1-3H3,(H2,57,68)(H,59,72)(H,60,73)(H,61,71)(H,62,69)(H,63,75)(H,64,70)(H,65,74);1H3,(H,3,4)/t30-,38-,40+,41+,42-,43+,44?,45+,46+;/m1./s1

InChI key

DEXPIBGCLCPUHE-HPDHVNDUSA-N

Application

Lanreotide acetate has been used as an inhibitor to test the plasticity of hydrogen sulfide modulation by growth hormone/thyroid hormone signaling in wild-type mice.

Biochem/physiol Actions

Lanreotide acetate is a somatostatin analog, a selective agonist for the SRIF-1 sst2 subtype of somatostatin receptor with a binding affinity of 0.8 nM for sst2 compared to 5.2 nM for sst5, 100 nM for sst3 and more than 1000 nM for the SRIF-2 subtypes, sst1 and sst4 receptors. It is used clinically in the management of acromegaly and symptoms caused by neuroendocrine tumors, and in recent studies can also inhibit tumor growth and has shown activity against non-endocrine tumors.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Sidéris et al.
The oncologist, 17(6), 747-755 (2012-05-26)
For decades, somatostatin analogs (including octreotide and lanreotide) have been indicated for relief of the symptoms of flushing, diarrhea, and wheezing associated with secretory neuroendocrine tumors (NETs). Recently, it has been suggested that somatostatin analogs may provide direct and indirect
Nancy M Gardner-Roehnelt
Clinical journal of oncology nursing, 16(1), 56-64 (2012-02-03)
Although neuroendocrine tumors (NETs) have been recognized as a family of complex malignancies since 1907, major progress has been made only in the past 20 years in understanding and managing the disease. The detection and reported incidence of NETs have
Christos Toumpanakis et al.
Seminars in oncology, 40(1), 56-68 (2013-02-09)
Somatostatin analogs (SA) are the standard of care for controlling symptoms of patients with functional gastro-entero-pancreatic neuroendocrine tumors (GEP-NETs). SA control symptoms in more than 70% of patients with carcinoid syndrome. Similar results are obtained in patients with functional, hormone-secreting
Ivan Borbath et al.
Acta gastro-enterologica Belgica, 75(2), 270-273 (2012-08-09)
We describe the case of a 54 years old woman, with hepatitis B, in whom the diagnosis of a 6 cm hepatocellular carcinoma (HCC) in the left liver was made in 2001. Alpha-foeto-protein (AFP) was 63 ng/mL (NI < 10
Shaun Coughlin et al.
World journal of surgery, 36(5), 1016-1029 (2012-03-16)
Enterocutaneous fistulas are abnormal connections between the skin and gastrointestinal tract that most commonly occur after surgery. Somatostatin analogues have been used in their treatment. The objective of the present study was to determine if somatostatin analogues shorten the time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service