Skip to Content
Merck
All Photos(1)

Key Documents

SML1586

Sigma-Aldrich

Oritavancin diphosphate

≥97% (HPLC)

Synonym(s):

LY333328 diphosphate, (4"R)-22-O-(3-Amino-2,3,6-trideoxy-3-C-methyl-α-L-arabino-hexopyranosyl)-N3"-[(4′-chloro[l,1′-biphenyl]-4-yl)methyl]vancomycin diphosphate, LY-333328

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C86H97Cl3N10O26 · 2H3PO4
CAS Number:
Molecular Weight:
1989.09
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[P](=O)(O)(O)O.[P](=O)(O)(O)O.Clc1ccc(cc1)c2ccc(cc2)CN[C@@]3([C@H]([C@@H](O[C@H](C3)O[C@H]4[C@@H](O[C@@H]([C@H]([C@@H]4O)O)CO)Oc5c6cc7cc5Oc8c(cc(cc8)[C@H]([C@H](C(=O)N[C@H](C(=O)N[C@H]7C(=O)N[C@@H]9c%10cc(c(cc%10)O)c%11c(cc(cc%11O)O)[C@H](NC(=O)[C@@H](NC9

InChI

1S/C86H97Cl3N10O26.2H3O4P/c1-35(2)22-51(92-7)77(110)98-67-69(105)42-15-20-55(49(88)24-42)120-57-26-44-27-58(73(57)125-84-74(71(107)70(106)59(34-100)122-84)124-62-32-86(6,76(109)37(4)119-62)93-33-38-8-10-39(11-9-38)40-12-17-45(87)18-13-40)121-56-21-16-43(25-50(56)89)72(123-61-31-85(5,91)75(108)36(3)118-61)68-82(115)97-66(83(116)117)48-28-46(101)29-54(103)63(48)47-23-41(14-19-53(47)102)64(79(112)99-68)96-80(113)65(44)95-78(111)52(30-60(90)104)94-81(67)114;2*1-5(2,3)4/h8-21,23-29,35-37,51-52,59,61-62,64-72,74-76,84,92-93,100-103,105-109H,22,30-34,91H2,1-7H3,(H2,90,104)(H,94,114)(H,95,111)(H,96,113)(H,97,115)(H,98,110)(H,99,112)(H,116,117);2*(H3,1,2,3,4)/t36-,37-,51+,52-,59+,61-,62-,64+,65+,66-,67+,68-,69+,70+,71-,72+,74+,75-,76-,84-,85-,86-;;/m0../s1

InChI key

PWTROOMOPLCZHB-BHYQHFGMSA-N

Related Categories

Biochem/physiol Actions

Oritavancin is a lipoglycopeptide vancomycin analog with broad spectrum activity against gram-positive pathogens, including methicillin-resistant Staphylococcus aureus (MRSA) and organisms resistant to vancomycin and other antibiotics such as linezolid and daptomycin. Oritavancin has multiple mechanisms of action, including inhibition of transglycosylation, inhibition of transpeptidation, and cell membrane interaction/disruption. Oritavancin also has a long half-life, allowing for a single intravenous dose of rather than the standard vacomycin treatment of twice-daily infusions for ten days.
Oritavancin is a lipoglycopeptide vancomycin analog with broad spectrum activity against gram-positive pathogens.

Caution

category 1 skin sensitizer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service