Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501896

Sigma-Aldrich

Anti-MMP-9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

92 kDa gelatinase, 92 kDa type IV collagenase precursor, CLG4B, GELB, Gelatinase B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

Related Categories

General description

Anti-MMP-9 Antibody detects endogenous levels of total MMP-9 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MMP-9.

Immunogen Range: 651-700

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Ge et al.
Molecular carcinogenesis, 62(9), 1399-1416 (2023-06-09)
Interleukin-17 (IL-17), a potent proinflammatory cytokine, can trigger the metastasis of non-small cell lung cancer (NSCLC). However, the underlying mechanism involved in IL-17-induced NSCLC cell metastasis remains unclear. In this study, we found that not only the expression of IL-17
Pengyu Liu et al.
OncoTargets and therapy, 13, 4099-4110 (2020-06-05)
This study aimed to investigate the regulatory role and mechanism of microRNA-766 (miR-766) on cutaneous squamous cell carcinoma (CSCC) cells. The expression of miR-766 and programmed cell death 5 (PDCD5) was detected in CSCC tissues and CSCC cell lines (A431
Li Duan et al.
PloS one, 8(9), e73768-e73768 (2013-09-17)
While aberrant activation of microglial cells was evidently involved in neuroinflammation and neurotoxicity in the neurodegenerative diseases such as Alzheimer's and Parkinson's disease, objective of study was to address if activated microglias deliver their effect by releasing pro-neurotrophins. By in
Fanlu Meng et al.
Experimental cell research, 374(2), 315-322 (2018-12-12)
Lung cancer is the leading cause of cancer-related death worldwide. Previous studies revealed that miR-183-5p is frequently involved in various human cancers. However, the exact role of miR-183-5p in regulating the pathogenesis of lung cancer remains unclear. Bioinformatic analysis, luciferase
Chen Liu et al.
Molecular medicine reports, 15(5), 3210-3214 (2017-03-30)
Overexpression of peptidyl-prolyl cis/trans isomerase, NIMA interacting‑1 (Pin1) is a significant marker of the occurrence and development of tumors. In the present study, the imidazoline ketone herbicide imazamethabenz was investigated as a potential antitumor drug by inhibiting Pin1. Molecular docking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service