Skip to Content
Merck
All Photos(4)

Key Documents

AV33090

Sigma-Aldrich

Anti-MMP9 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

78 kDa

species reactivity

human, dog, mouse, guinea pig, horse, bovine, sheep, rat, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

Related Categories

General description

Matrix metalloproteinase-9/gelatinase B (MMP-9), a member of the matrix metalloproteinase (MMP) family, is involved in the breakdown of extracellular matrix giving it a role in normal physiological processes such as embryonic development, tissue remodeling, wound repair, vascularization and inflammatory response. MMP-9 is involved in synaptic plasticity. MMP-9 functions in modulating hippocampal synaptic physiology and plasticity.

Immunogen

Synthetic peptide directed towards the N terminal region of human MMP9

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Rabbit polyclonal anti-MMP9 antibody is used to tag matrix metalloproteinase-9/gelatinase B for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of matrix metalloproteinase-9/gelatinase B in processe such as synaptic plasticity.

Biochem/physiol Actions

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by MMP9 degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling.

Sequence

Synthetic peptide located within the following region: VAEHGDGYPFDGKDGLLAHAFPPGPGIQGDAHFDDDELWSLGKGVVVPTR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashant Nighot et al.
American journal of physiology. Gastrointestinal and liver physiology, 309(12), G988-G997 (2015-10-31)
Recent studies have implicated a pathogenic role for matrix metalloproteinases 9 (MMP-9) in inflammatory bowel disease. Although loss of epithelial barrier function has been shown to be a key pathogenic factor for the development of intestinal inflammation, the role of
Gokhan Gundogdu et al.
International journal of impotence research, 36(3), 269-274 (2023-02-14)
Peyronie's disease (PD) is a debilitating pathology which is associated with penile curvature and erectile dysfunction due to the formation of fibrotic plaques in the penile tunica albuginea. In the present study, we developed a novel rabbit model of PD
Perle Latré De Laté et al.
Cellular microbiology, 21(3), e12973-e12973 (2018-11-10)
Constitutive c-Jun N-terminal kinase (JNK) activity characterizes bovine T and B cells infected with Theileria parva, and B cells and macrophages infected with Theileria annulata. Here, we show that T. annulata infection of macrophages manipulates JNK activation by recruiting JNK2 and
K Nagaoka et al.
Oncogene, 35(22), 2893-2901 (2015-09-29)
In mouse mammary epithelial cells, cytoplasmic polyadenylation element binding protein 1 (CPEB1) mediates the apical localization of ZO-1 mRNA, which encodes a critical tight junction component. In mice lacking CPEB1 and in cultured cells from which CPEB has been depleted
Qiang Zhang et al.
Chinese medical journal, 135(9), 1076-1086 (2022-02-23)
Fibrosis in the peripheral airways contributes to airflow limitation in patients with chronic obstructive pulmonary disease (COPD). However, the key proteins involved in its development are still poorly understood. Thus, we aimed to identify the differentially expressed proteins (DEPs) between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service