Skip to Content
Merck
All Photos(2)

Key Documents

AB19016

Sigma-Aldrich

Anti-MMP-9 Antibody, Catalytic domain

Chemicon®, from rabbit

Synonym(s):

Gelatinase B, 92 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

General description

All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell matrix interactions. MMPs are Zn(2+) binding endopeptidases that degrade various components of the ECM.
MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. MMP9, also known as gelatinase B, is a secreted enzyme which degrades the interstitial collagens, types I, II, and III, and is produced by normal alveolar macrophages and granulocytes. The expression of MMP9 increases in Epstein-Barr virus infected lymphoma derived cell lines and may be of significance in typically invasive nasopharyngeal carcinomas.

Specificity

Reactivity with other species has not been confirmed.
Recognizes rat MMP-9 pro and active forms. Antibody also reacts with mouse MMP-9 and human MMP-9, other species not tested. Exhibits no cross-reactivity with other MMP family members. The antibody was generated using E. coli-expressed active rat 92 kDa type IV collagenase (catalytic domain) as an immunogen.

Immunogen

E. coli-expressed active rat 92 kDa type IV collagenase (catalytic domain)
Epitope: Catalytic domain

Application

Anti-MMP-9 Antibody, Catalytic domain is an antibody against MMP-9 for use in IH, IP & WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blot Analysis:
1:1000 dilution of a previous lot was used.

Immunoprecipitation:
1:500 dilution from a previous lot was used. MMP-9 is constitutively produced by some tumor cell lines (i.e., HT1080, HL60, and U937), but not in most quiescent cells and tissues. The phorbol ester TPA stimulates production of MMP-9 in some cell types. MMP-9 levels in culture media (pg/mL) are often below the threshold of detection by standard Western Blotting. The enzyme can be concentrated from culture media by gelatin-agarose affinity chromatography (Goldberg, G.I. et al. J. Biol. Chem. 267, 4583-4591. 1992). This can prevent appearance of a spurious band of antibody binding in the vicinity of 68 kDa displayed by some concentrated media.
Western blot: 1:2,000. Typically 98-95 kDa pro and 92-88kDa active forms are detected as well as breakdown products under fully reduced conditions. Under non-reduced conditions additional bands may be detected because of SDS-stable TIMP and other inhibitor complexes. CC069 is recommended as a positive control.

Immunohistochemistry: 1:100 dilution from a previous lot was used in frozen acetone fixed tissues.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on L6 lysates.

Western Blot Analysis: 1:1000 dilution of this lot detected MMP-9 on 10 µg of L6 lysates.

Target description

92 kDa

Linkage

Replaces: 04-1150

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing PBS, containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HL60 cell lysate, U937 cell lysate Lung.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasminogen is a joint-specific positive or negative determinant of arthritis pathogenesis in mice.
Raghu, H; Jone, A; Cruz, C; Rewerts, CL; Frederick, MD; Thornton, S; Degen, JL; Flick, MJ
Arthritis & Rheumatology (Hoboken, N.J.) null
Fei Yu et al.
Molecular medicine reports, 15(5), 2969-2976 (2017-03-31)
ORMDL sphingolipid biosynthesis regulator 3 (ORMDL3) has been previously implicated in asthma pathogenesis, its effect on airway remodeling remains to be elucidated. The present study examined the expression levels of ORMDL3 in a mouse model of asthma. Mice were divided into
Le Thi Anh Hong et al.
Nature communications, 8(1), 533-533 (2017-09-16)
The cystic cavity that develops following injuries to brain or spinal cord is a major obstacle for tissue repair in central nervous system (CNS). Here we report that injection of imidazole-poly(organophosphazenes) (I-5), a hydrogel with thermosensitive sol-gel transition behavior, almost
Aparna Karthikeyan et al.
Oncotarget, 9(38), 24950-24969 (2018-06-05)
Glioma tumors constitute a significant portion of microglial cells, which are known to support tumor progression. The present study demonstrates that transforming growth factor-β (TGFβ) signaling pathway in microglia in a glioma environment is involved in tumor progression and pathogenesis.
Ana Peñalver et al.
Frontiers in molecular neuroscience, 10, 278-278 (2017-09-21)
Lysophosphatidic acid (LPA) is an extracellular lipid mediator that regulates nervous system development and functions acting through G protein-coupled receptors (GPCRs). Here we explore the crosstalk between LPA1 receptor and glutamatergic transmission by examining expression of glutaminase (GA) isoforms in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service