Skip to Content
Merck
All Photos(1)

Key Documents

RAB1169

Sigma-Aldrich

Human GSTP1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GSTP1(2950)

General description

This ELISA antibody pair detects human GSTP1. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria-Cristina Arcangeletti et al.
International journal of molecular sciences, 21(17) (2020-09-10)
Systemic sclerosis (SSc) is a severe autoimmune disorder characterized by vasculopathy and multi-organ fibrosis; its etiology and pathogenesis are still largely unknown. Herpesvirus infections, particularly by human cytomegalovirus (HCMV) and human herpesvirus 6 (HHV-6), have been suggested among triggers of
Marc Bourouis et al.
PloS one, 14(2), e0209759-e0209759 (2019-02-09)
Basal autophagy is as a compressive catabolic mechanism engaged in the breakdown of damaged macromolecules and organelles leading to the recycling of elementary nutrients. Thought essential to cellular refreshing, little is known about the origin of a constitutional rate of
Michael G Leitner et al.
Cellular and molecular life sciences : CMLS (2018-07-11)
PTEN prevents tumor genesis by antagonizing the PI3 kinase/Akt pathway through D3 site phosphatase activity toward PI(3,4)P2 and PI(3,4,5)P3. The structural determinants of this important specificity remain unknown. Interestingly, PTEN shares remarkable homology to voltage-sensitive phosphatases (VSPs) that dephosphorylate D5
Tao Guo et al.
Plant physiology, 182(3), 1346-1358 (2019-12-29)
Phosphoinositides (PIs) as regulatory membrane lipids play essential roles in multiple cellular processes. Although the exact molecular targets of PI-dependent modulation remain largely elusive, the effects of disturbed PI metabolism could be employed to identify regulatory modules associated with particular
Ying Yang et al.
The Journal of clinical endocrinology and metabolism, 104(10), 4341-4346 (2019-05-11)
Evidence indicates that there is substantial impairment/loss of β-cell function/mass even before prediabetes. Elevated plasma proinsulin is a sign of β-cell dysfunction in patients with diabetes/prediabetes. However, the dynamic changes of glucose stimulated proinsulin secretion (GSPS) among nondiabetic individuals remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service