Skip to Content
Merck
All Photos(3)

Key Documents

16337

Supelco

Fucoxanthin

analytical standard

Synonym(s):

all-trans-Fucoxanthin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H58O6
CAS Number:
Molecular Weight:
658.91
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(=O)O[C@H]1CC(C)(C)C(=C=C\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(/C)C(=O)C[C@@]23O[C@]2(C)C[C@@H](O)CC3(C)C)[C@](C)(O)C1

InChI

1S/C42H58O6/c1-29(18-14-19-31(3)22-23-37-38(6,7)26-35(47-33(5)43)27-40(37,10)46)16-12-13-17-30(2)20-15-21-32(4)36(45)28-42-39(8,9)24-34(44)25-41(42,11)48-42/h12-22,34-35,44,46H,24-28H2,1-11H3/b13-12+,18-14+,20-15+,29-16+,30-17+,31-19+,32-21+/t23-,34-,35-,40+,41+,42-/m0/s1

InChI key

SJWWTRQNNRNTPU-XJUZQKKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fucoxanthin, an abundant marine xanthophyll and a type of sea-weed carotenoid, plays a key role in generating the brown pigment in brown seaweeds. It exhibits pharmacological properties including antitumor and antioxidant properties.

Application

Fucoxanthin may be used as an analytical standard for the quantification of the analyte in Undaria pinnatifida using reversed-phase high-performance liquid chromatography (RP-HPLC) technique. Its potential as a selective anticancer agent and an active component for the anti-obesity effect in Undaria lipids is investigated.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Xanthophyll carotenoid pigment extracted from algae. Exhibits anticancer, antioxidant, anti-obesity and anti-inflammatory properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Food Factors for Health Promotion (2009)
Fucoxanthin from edible seaweed, Undaria pinnatifida, shows antiobesity effect through UCP1 expression in white adipose tissues
Maeda H, et al.
Biochemical and Biophysical Research Communications, 332(2), 392-397 (2005)
Extracts from New Zealand Undaria pinnatifida containing fucoxanthin as potential functional biomaterials against cancer in vitro
Wang KS, et al.
Journal of Functional Biomaterials, 5(2), 29-42 (2014)
Ga-Young Seo et al.
International journal of molecular sciences, 20(3) (2019-01-31)
Leathesia difformis (L.) Areschoug (L. difformis) is a species of littoral brown algae of the class Phaeophyceae. Only a few studies on the apoptotic, antiviral, and antioxidant properties of L. difformis have been reported, and its inhibitory effect on melanin
Sangeetha Ravi Kumar et al.
Marine drugs, 11(12), 5130-5147 (2013-12-20)
Fucoxanthin is a marine carotenoid exhibiting several health benefits. The anti-cancer effect of fucoxanthin and its deacetylated metabolite, fucoxanthinol, is well documented. In view of its potent anti-carcinogenic activity, the need to understand the underlying mechanisms has gained prominence. Towards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service