Skip to Content
Merck
All Photos(2)

Key Documents

RAB0232

Sigma-Aldrich

Human IGFBP-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGFBP1(3484)

General description

The Human IGF-BP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IGF-BP-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IGFBP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subeen Hong et al.
Archives of gynecology and obstetrics, 301(6), 1431-1439 (2020-05-06)
To investigate whether complement and other immune-related proteins in cervicovaginal fluid (CVF) can predict intra-amniotic infection and/or inflammation (IAI) and spontaneous preterm delivery (SPTD, < 34.0 weeks) in women with preterm labor (PTL) and to compare the predictive abilities of these biomarkers with
Giovanni Sisti et al.
Medicina (Kaunas, Lithuania), 55(5) (2019-05-19)
Background and Objectives: To investigate if pregnancies conceived using an oocyte donor necessitate an alteration in immune regulation, we compared concentrations of insulin-like growth factor binding protein (IGFBP)-1, insulin-like growth factor (IGF)-1 and T cell immunoglobulin mucin-3 (Tim-3) in women
Takumi Yamane et al.
Bioscience, biotechnology, and biochemistry, 83(3), 511-517 (2018-11-13)
Although starvation has been reported to influence the functions of various tissues, its effects on the skin are not well understood. In this study, we investigated the effect of starvation on hyaluronan synthesis in rat skin. Starvation reduced hyaluronan synthesis
Pavel Deryabin et al.
Frontiers in cell and developmental biology, 9, 624053-624053 (2021-03-05)
Endometrium is the uterine lining that undergoes hundreds of cycles of proliferation, differentiation, and desquamation throughout a woman's reproductive life. Recently, much attention is paid to the appropriate endometrial functioning, as decreased endometrial receptivity is stated to be one of
Giovanni Sisti et al.
Archives of gynecology and obstetrics, 300(3), 583-587 (2019-06-16)
Mechanisms leading to preterm premature rupture of membranes (PPROM) remain incompletely defined. Based on the elevated occurrence of PPROM in twin gestations and recent studies of the involvement of insulin-like growth factor binding protein-1 (IGFBP-1) in the inhibition of collagen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service