Skip to Content
Merck
All Photos(4)

Key Documents

HAWG047S6

Millipore

S-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis

Synonym(s):

Membrane filters, Semi-permeable membranes, S-Pak Filters 0.45µm 47mm white gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
white filter

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water

manufacturer/tradename

S-Pak®

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample type beverage(s)
sample type cider
sample type food(s)
sample type sport drink
sample type soft drink
sample(s) (environmental)
sample type cosmetics
sample type beer
sample type bottled water
sample type wine

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

S-Pak Filters are made from Mixed cellulose Esters (MCE) with 0.45 μm pore size, white, gridded, 47mm diameter. These sterile, ready to use, individually packed filters are easy to use with high recoveries and more appropriate for general microbiological testings.

Application

The filters have been manufactured for use in the membrane filtration method of bacterial analysis of water and beverages such as water, wine, beer, sport drinks. They are also used in analysis of pharmaceutical and cosmetic products.

Packaging

sealed, with blue separator paper, sterile

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franciska M Schets et al.
Environment international, 137, 105516-105516 (2020-02-03)
Swimming ponds are artificial ecosystems for bathing in which people imitate the conditions of natural waters. Swimming in natural water may pose health risks if the water quality is microbiologically poor. Swimming ponds are small water bodies that may be
Jasha Dehm et al.
Marine pollution bulletin, 156, 111239-111239 (2020-06-09)
Microplastics (MPs) remain largely understudied in Small Island Developing States. This study is aimed at comparing the abundance and characteristics of MPs in rural and urban marine coastal sites located along the southern coast of Fiji's main inhabited island, Viti
Fuhar Dixit et al.
Journal of hazardous materials, 384, 121261-121261 (2019-10-02)
Carcinogenic GenX chemicals, heptafluoropropylene-oxide-dimer-acid (HFPO-DA), have been recently detected in surface, ground and recycled water sources worldwide. However, GenX removals under the influence of variable characteristics of the organic and inorganic compounds present in the natural water sources, have often

Articles

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

Related Content

S-Pak® membrane filters for microbial bioburden testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service