Skip to Content
Merck
All Photos(1)

Key Documents

SAB3600001

Sigma-Aldrich

Anti-IGF-1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Insulin-like Growth Factor-I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

lyophilized powder

species reactivity

human

technique(s)

immunocytochemistry: 5-15 μg/mL
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 0.1-0.2 μg/mL

application(s)

research pathology

storage temp.

−20°C

Gene Information

human ... IGF1(3479)

General description

Insulin-like growth factor-I (IGF-I) is encoded by the gene mapped to human chromosome 12q23.2. IGF-I belongs to insulin-like growth factor (IGF) family. The encoded protein is ubiquitously expressed, but the circulating form of the protein is mainly found in liver and pancreas.

Immunogen

purified, E. coli-derived recombinant human insulin-like growth factor-I.

Application

Anti- IGF-1 antibody may be used for neutralization reactions, the ND50 of the antibody is 3-12 μg/ml. For immunoblotting a working antibody concentration of 0.1-0.2 μg/ml. The recommended antibody concentration for immunocytochemistry is 5-15 μg/ml. The antibody is suitable for immunohistochemistry of formalin-fixed, paraffin-embedded sections.

Biochem/physiol Actions

Insulin-like growth factors -1 and -2 are important regulators of signalling that mediates growth and metabolism in cells. Insulin-like growth factor -1 shares structural homology with proinsulin and is mainly produced by liver. IGF-1 acts via specific receptor, IGF-1R, a heterodimeric tyrosine kinase receptor that signals to various second messenger pathways such as MAPK, Raf/Ras and PI3K cascades. The effect of IGF-1 is by endocrine, paracrine and autocrine signalling. The effects of IGF-1 signalling are important at many developmental levels from cells to organ to organism, growth, differentiation, anti-apoptosis, proliferation and transformation. Irregular IGF-1 signaling has been implicated in many cancers including melanoma, breast, lung, colon and prostate. Anti-IGF-I recognizes human insulin-like growth factor I.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-like growth factor system and sporadic malignant melanoma.
Capoluongo E et al
The American Journal of Pathology, 178, 26-31 (2011)
Insulin-like growth factor 1 (IGF-1) expression is up-regulated in lymphoblastoid cell lines of lithium responsive bipolar disorder patients.
Squassina A, et al.
Pharmacological Research, 73, 1-7 (2013)
H Yu et al.
Journal of the National Cancer Institute, 91(2), 151-156 (1999-01-29)
Insulin-like growth factors (IGFs), in particular IGF-I and IGF-II, strongly stimulate the proliferation of a variety of cancer cells, including those from lung cancer. To examine the possible causal role of IGFs in lung cancer development, we compared plasma levels
J M Chan et al.
Science (New York, N.Y.), 279(5350), 563-566 (1998-02-07)
Insulin-like growth factor-I (IGF-I) is a mitogen for prostate epithelial cells. To investigate associations between plasma IGF levels and prostate cancer risk, a nested case-control study within the Physicians' Health Study was conducted on prospectively collected plasma from 152 cases
J Dupont et al.
Hormone research, 55 Suppl 2, 22-26 (2001-10-31)
The insulin receptor (IR) and the insulin-like growth factor I receptor (IGF-IR) belong to the same subfamily of receptor tyrosine kinases with two extracellular alpha-subunits and two transmembrane beta-subunits. They share a high similarity of structure and intracellular signalling events.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service