RAB0223
Human IFN γ ELISA Kit
for cell and tissue lysates
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.77
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type tissue lysate
sample type cell lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 20.6-15000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IFNG(3458)
General description
The Human IFN-γ ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IFN-γ cell lysate and tissue lysate.
Immunogen
Recombinant Human IFN-γ
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human IFNγ ELISA Kit has been used in ELISA assay.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human IFNγ ELISA Kit has been used in ELISA assay.
Biochem/physiol Actions
IFNG (interferon-γ) is a type-2 interferon, which is produced by Th (T helper cells)-1 lymphocytes, CD8 cytotoxic lymphocytes, natural killer (NK) cells, and natural killer T (NKT) cells. It plays an important role in innate and adaptive immune responses, and helps in defense against bacterial infections, dimorphic yeast and some viral infections. High expression of IFNG is common in many autoimmune diseases, such as systemic lupus erythematosis, Sjögren syndrome, inflammatory bowel disease and multiple sclerosis.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Association between B7-H1 and cervical cancer: B7-H1 impairs the immune response in human cervical cancer cells.
Tao J
Experimental and Therapeutic Medicine, 14(5), 4125-4133 (2017)
Yong-Chen Lu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(13), 3401-3410 (2014-07-06)
Cancer immunotherapy with adoptive transfer of tumor-infiltrating lymphocytes (TIL) represents an effective treatment for patients with metastatic melanoma, with the objective regressions in up to 72% of patients in three clinical trials. However, the antigen targets recognized by these effective
Benjamin J Murdock et al.
Journal of immunology (Baltimore, Md. : 1950), 193(8), 4107-4116 (2014-09-17)
The potent immunoregulatory properties of IL-10 can counteract protective immune responses and, thereby, promote persistent infections, as evidenced by studies of cryptococcal lung infection in IL-10-deficient mice. To further investigate how IL-10 impairs fungal clearance, the current study used an
Samuel T Haile et al.
Cancer immunology research, 2(7), 610-615 (2014-05-14)
Tumor cells use various methods of immunosuppression to overcome antitumor immunity. One such method is that of programmed death ligand-1 (PD-L1 or B7-H1), which upon binding its receptor PD-1 on T cells triggers apoptotic death of the activated T cells.
Vibha Jha et al.
PloS one, 9(8), e104484-e104484 (2014-08-15)
Environmental factors including drugs, mineral oils and heavy metals such as lead, gold and mercury are triggers of autoimmune diseases in animal models or even in occupationally exposed humans. After exposure to subtoxic levels of mercury (Hg), genetically susceptible strains
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service