Skip to Content
Merck
All Photos(2)

Key Documents

RAB0225

Sigma-Aldrich

Mouse IFN-γ ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ifng(15978)

General description

The Mouse IFN-γ ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IFN-γ in cell lysate and tissue lysate.

Immunogen

Recombinant Mouse IFN-γ

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

K Kim et al.
Tuberculosis (Edinburgh, Scotland), 94(5), 494-501 (2014-08-07)
Previous studies suggest that control of Mycobacterium tuberculosis infection is compromised by the activity of regulatory T cells, including those that express CD39, an ectonucleotidase with immunosuppressive properties. Here, we examine the role of CD39 on CD4+ T cells reacting
Kaveh Abdi et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 6028-6036 (2014-05-14)
IL-12p40 partners with the p35 and p19 polypeptides to generate the heterodimeric cytokines IL-12 and IL-23, respectively. These cytokines play critical and distinct roles in host defense. The assembly of these heterodimers is thought to take place within the cell
Chris P Verschoor et al.
Infection and immunity, 82(11), 4824-4833 (2014-08-27)
Pneumonia caused by Streptococcus pneumoniae is a major cause of death and an economic burden worldwide. S. pneumoniae is an intermittent colonizer of the human upper respiratory tract, and the ability to control asymptomatic colonization determines the likelihood of developing
Stefanie Raab et al.
Journal of immunology (Baltimore, Md. : 1950), 193(8), 4261-4272 (2014-09-14)
The ability of NK cells to mediate Ab-dependent cellular cytotoxicity (ADCC) largely contributes to the clinical success of antitumor Abs, including trastuzumab, which is approved for the treatment of breast cancer with HER2/neu overexpression. Notably, only ∼25% of breast cancer
Xiaowei Wu et al.
Chemistry, an Asian journal, 9(8), 2260-2271 (2014-04-05)
We have previously described the discovery of N-alkylated iminosugars that showed immunosuppressive activity both in vitro and in vivo. Herein, we report the synthesis and biological evaluation of N-arylated lactam-type iminosugar derivatives. The synthesis started from simple monosaccharides and featured a Buchwald-Hartwig

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service