Skip to Content
Merck
All Photos(1)

Key Documents

R5131

Sigma-Aldrich

Anti-Rabbit Serum antibody produced in goat

whole antiserum

Synonym(s):

Anti-Serum, rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

rabbit

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Rabbit serum can be used for the study of serum proteins and antibody-mediated immune responses . Anti-Rabbit Serum antibody is specific for normal rabbit serum, and has not been assayed for cross reactivity with other species.

Specificity

Strong reactivity with normal rabbit serum has been determined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies crossreactivity.

Immunogen

Rabbit serum

Application

Anti-Rabbit Serum antibody is suitable for use in immunoelectrophoresis.
Anti-Rabbit Serum antibody produced in goat has been used in
  • incubation of rabbit blastocyst prior to inner cell mass
  • isolation
  • RNA isolation
  • western blotting

Physical form

Goat Anti-Rabbit Serum is provided as a liquid containing 0.1% sodium azide (see MSDS) as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

STUDIES ON HEAT-LABILE OPSONIN IN RABBIT SERUM.
J G HIRSCH et al.
Journal of immunology (Baltimore, Md. : 1950), 92, 145-154 (1964-01-01)
Differentiation of derived rabbit trophoblast stem cells under fluid shear stress to mimic the trophoblastic barrier
Sanz G, et al.
Biochim. Biophys. Acta Gen. Subj. (2019)
Guenhaël Sanz et al.
Biochimica et biophysica acta. General subjects, 1863(10), 1608-1618 (2019-07-07)
The placenta controls exchanges between the mother and the fetus and therefore fetal development and growth. The maternal environment can lead to disturbance of placental functions, with consequences on the health of the offspring. Since the rabbit placenta is very
Ethylene and carbon dioxide production by developing strawberries show a correlative pattern that is indicative of ripening climacteric fruit
Iannetta PPM , et al.
Physiologia Plantarum, 127(2), 247-259 (2006)
Eutherian mammals use diverse strategies to initiate X-chromosome inactivation during development
Okamoto I, et al.
Nature, 472(7343), 370-370 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service