Skip to Content
Merck
All Photos(2)

Key Documents

1.06007

Supelco

Methanol

≥99.9% (GC), gradient grade, suitable for HPLC, LiChrosolv®, reag. Ph. Eur.

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NA.03
Assay:
≥99.9% (GC)
technique(s):
HPLC: suitable
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur

grade

gradient grade
gradient grade

Quality Level

Agency

reag. Ph. Eur.

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤2.0 mg/L

color

APHA: ≤10

transmittance

210 nm, ≥20%
220 nm, ≥60%
230 nm, ≥75%
235 nm, ≥83%
250 nm, ≥95%
260 nm, ≥98%

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

UV absorption

λ: 225 nm Amax: ≤0.170

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application

Methanol is used as a solvent:
  • In the mobile phase to separate mercury species on a Zorbax C18 reverse phase column. [1]
  • To wash the cartridges and as an eluent to extract the analytes during the solid-phase extraction (SPE) process. [2]

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.02 %
Color: ≤ 10 Hazen
Density (d 20 °C/20 °C): 0.791 - 0.793
Boiling point: 64 - 65 °C
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Gradient grade (at 235 nm): ≤ 2.0 mAU
Gradient grade (at 254 nm): ≤ 1.0 mAU
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 0.5 ppb
Transmission (at 210 nm): ≥ 20 %
Transmission (at 220 nm): ≥ 60 %
Transmission (at 230 nm): ≥ 75 %
Transmission (at 235 nm): ≥ 83 %
Transmission (at 250 nm): ≥ 95 %
Transmission (from 260 nm): ≥ 98 %
Absorbance (at 225 nm): ≤ 0.170
Filtered by 0.2 µm filter.
Suitable for UPLC / UHPLC /Ultra HPLC - instruments.
Conforms to Methanol R1 and R2 according to Reag. Ph Eur;
conforms to the requirements of ACS for liquid chromatography suitability.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of mercury species by the diffusive gradient in thin film technique and liquid chromatography--atomic fluorescence spectrometry after microwave extraction.
Pavlna P et al.
Analytica Chimica Acta, 866, 21-26 (2015)
Heterocyclic amines in aged and thermally treated pork longissimus dorsi muscle of normal and PSE quality.
T Polak et al.
LWT--Food Science and Technology, 42, 504-513 (2009)

Articles

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

See All

Protocols

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service