Skip to Content
Merck
All Photos(1)

Key Documents

P9248

Sigma-Aldrich

PD 169316

≥98% (HPLC), solid

Synonym(s):

4-(4-Fluorophenyl)-2-(4-nitrophenyl)-5-(4-pyridyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13FN4O2
CAS Number:
Molecular Weight:
360.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

faint yellow to dark orange

solubility

DMSO: >10 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(cc1)-c2nc(-c3ccc(F)cc3)c([nH]2)-c4ccncc4

InChI

1S/C20H13FN4O2/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(8-4-15)25(26)27/h1-12H,(H,23,24)

InChI key

BGIYKDUASORTBB-UHFFFAOYSA-N

Gene Information

human ... MAPK14(1432)

Related Categories

General description

PD 169316 is a pyridinyl imidazole compound. It is a potential inhibitor of p38 mitogen-activated protein kinases. It also inhibits signalling transforming growth factor β (TGFβ), particularly in human ovarian cancer cells.

Application

PD 169316 has been used for the inhibition of p38 enzyme in human hepatocytes. It has been used in culture media to promote embryoid bodies differentiation.

Biochem/physiol Actions

Potent, cell-permeable and selective p38 MAP kinase inhibitor (IC50 = 89 nM).

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of CD40 with platelet derived CD154 promotes reactive oxygen species dependent death of human hepatocytes during hypoxia and reoxygenation
Bhogal RH, et al.
PLoS ONE, 7(1), e30867-e30867 (2012)
Inhibition of p38 kinase mimics survival signal-linked protection against apoptosis in rat cerebellar granule neurons
Nath R, et al.
Cellular & Molecular Biology Letters, 6(2), 173-184 (2001)
A p38MAPK-p53 cascade regulates mesodermal differentiation and neurogenesis of embryonic stem cells
Hadjal Y, et al.
Cell Death & Disease, 4(7), e737-e737 (2013)
The p38 MAPK inhibitor, PD169316, inhibits transforming growth factor beta-induced Smad signaling in human ovarian cancer cells
Fu Y, et al.
Biochemical and Biophysical Research Communications, 310(2), 391-397 (2003)
Byoung Kwon Park et al.
Biomolecules & therapeutics, 29(3), 273-281 (2021-01-29)
Severe acute respiratory syndrome CoV-2 (SARS-CoV-2) is responsible for the current coronavirus disease 2019 (COVID-19) pandemic. Signaling pathways that are essential for virus production have potential as therapeutic targets against COVID-19. In this study, we investigated cellular responses in two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service