Skip to Content
Merck
All Photos(1)

Key Documents

U120

Sigma-Aldrich

U0126 ethanolate

≥98% (HPLC), powder

Synonym(s):

1,4-Diamino-2,3-dicyano-1,4-bis(o-aminophenylmercapto)butadiene ethanolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N6S2 · C2H5OH
CAS Number:
Molecular Weight:
426.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to off-white

solubility

DMSO: >10 mg/mL (Stock solutions in DMSO should be used within a month. Acidic solutions exhibit weaker inhibition)

storage temp.

2-8°C

SMILES string

N#CC(C(C#N)=C(N)SC1=C(N)C=CC=C1)=C(N)SC2=C(N)C=CC=C2.CCO

InChI

1S/C18H16N6S2.C2H6O/c19-9-11(17(23)25-15-7-3-1-5-13(15)21)12(10-20)18(24)26-16-8-4-2-6-14(16)22;1-2-3/h1-8H,21-24H2;3H,2H2,1H3

InChI key

CFQULUVMLGZVAF-UHFFFAOYSA-N

Gene Information

Application

U0126 ethanolate has been used as an MEK inhibitor in mononucleated myogenic cells derived from breast muscles of chicken embryos. U0126 has also been used to study ERK2 function in amyloid precursor protein (APP) knockout (KO) mice neurons.

Biochem/physiol Actions

U0126, 1,4-diamino-2,3-dicyano-1,4-bis[2-aminophenylthio] butadiene, is a highly selective inhibitor of both MEK1 and MEK2. U0126 was found to functionally antagonize AP-1 transcriptional activity via noncompetitive inhibition of the dual specificity kinase MEK with IC50 of 72 nM for MEK1 and 58 nM for MEK2. U0126 inhibited anchorage-independent growth of Ki-ras-transformed rat fibroblasts by simultaneously blocking both extracellular signal-regulated kinase (ERK) and mammalian target of rapamycin (mTOR)-p70(S6K) pathways. The effects of U0126 on the growth of eight human breast cancer cell lines shown that U0126 selectively repressed anchorage-independent growth of MDA-MB231 and HBC4 cells, two lines with constitutively activated ERK. Loss of contact with substratum triggers apoptosis in many normal cell types, a phenomenon termed anoikis. U0126 sensitized MDA-MB231 and HBC4 to anoikis, i.e., upon treatment with U0126, cells deprived of anchorage entered apoptosis.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

photosensitive

Preparation Note

U0126 ethanolate is soluble in DMSO at a concentration >10 mg/ml. Stock solutions of the product in DMSO should be used within a month. Acidic solutions of the product exhibit weaker MEK inhibitory functions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Xiao et al.
Cellular and molecular gastroenterology and hepatology, 8(1), 21-36 (2019-03-05)
Regenerating islet-derived protein (REG3γ), an antimicrobial peptide, typically expressed by intestinal epithelial cells (IEC), plays crucial roles in intestinal homeostasis and controlling gut microbiota. However, the mechanisms that regulate IEC expression of REG3γ are still largely unclear. In this study
Xiao-Yang Wang et al.
Molecular and cellular biology, 28(11), 3589-3599 (2008-03-26)
The RNA-binding protein Musashi1 (Msi1) is a positive regulator of Notch-mediated transcription in Drosophila melanogaster and neural progenitor cells and has been identified as a putative human breast stem cell marker. Here we describe a novel functional role for Msi1:
D R DeSilva et al.
Journal of immunology (Baltimore, Md. : 1950), 160(9), 4175-4181 (1998-05-09)
Three mitogen-activated protein kinase pathways are up-regulated during the activation of T lymphocytes, the extracellular signal-regulated kinase (ERK), Jun NH2-terminal kinase, and p38 mitogen-activated protein kinase pathways. To examine the effects of blocking the ERK pathway on T cell activation
Michael G Kharas et al.
The Journal of clinical investigation, 118(9), 3038-3050 (2008-08-16)
Some cases of pre-B cell acute lymphoblastic leukemia (pre-B-ALL) are caused by the Philadelphia (Ph) chromosome-encoded BCR-ABL oncogene, and these tend to have a poor prognosis. Inhibitors of the PI3K/AKT pathway reduce BCR-ABL-mediated transformation in vitro; however, the specific PI3K
Alessandra Ciullo et al.
International journal of molecular sciences, 20(3) (2019-01-27)
Cell therapy has been evaluated to enhance heart function after injury. Delivered cells mostly act via paracrine mechanisms, including secreted growth factors, cytokines, and vesicles, such as exosomes (Exo). Intramyocardial injection of cardiac-resident progenitor cells (CPC)-derived Exo reduced scarring and

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service