Skip to Content
Merck
All Photos(1)

Key Documents

P9208

Sigma-Aldrich

Polymyxin Acylase from Pseudomonas sp.

≥0.1 units/mg solid

Synonym(s):

Peptide N-fatty acylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Pseudomonas spp.)

Quality Level

form

solid

specific activity

≥0.1 units/mg solid

relevant disease(s)

cancer

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Polymyxin acylase, from Pseudomonas sp., is a N-myristoyl cleaving enzyme that has been used to determine the N-myristoyl peptide sequence and may be useful in cancer research since it has antitumor activity against murine and human tumor cells .

Biochem/physiol Actions

Polymyxin acylase, from Pseudomonas sp. deacylates polymyxin group antibiotics and long-chain fatty acyl groups of proteins. Polymyxin acylase has an affinity for long-chain fatty acyl proteins in human carcinoma cells .

Quality

Crude actone powder;

Unit Definition

One unit will hydrolyze 1 μmole of N-octanoyl-5-aminovaleric acid per min at pH 8.0 at 37°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Misumi et al.
Biochemical and biophysical research communications, 217(2), 632-639 (1995-12-14)
Polymyxin acylase isolated from Pseudomonas sp. M-6-3 was used as an N-myristoyl cleaving enzyme in order to determine a part of the N-terminal amino acid sequence of N-myristoyl proteins. The enzyme hydrolyzed a number of N-myristoyl oligopeptides at various hydrolysis
N Yasuda et al.
Biological & pharmaceutical bulletin, 18(4), 615-617 (1995-04-01)
Polymyxin acylase from Pseudomonas sp. M-6-3 can deacylate not only polymyxin group antibiotics, but also the long-chain fatty acyl group of proteins and peptides. We found the in vitro antitumor activity of polymyxin acylase against murine and human tumor cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service