01818
Acylase I from Aspergillus melleus
powder, brown, >0.5 U/mg
Synonym(s):
Acylase ‘Amano’, Aminoacylase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
Acylase I belongs to the aminoacylase family of enzymes.
Application
Acylase I from Aspergillus melleus has been used to catalyze the Mannich reaction.
Biochem/physiol Actions
Acylase I plays an important role in the amino acid metabolism of organisms. It also plays a role in the acylation of primary and secondary alcohols. Acylase I is involved in alcoholysis.
Unit Definition
1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol N-acetyl-L-methionine per minute at pH 8.0 and 37°C
Analysis Note
Enzyme activity: the optimum temperature is 40-45°C, the optimum pH is 8.0 (stable form pH 6-10). The enzyme is activated by CoCl2 in the range of 10-4 to 10-3 M.
Other Notes
Resolution of acetyl amino acids
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of Labelled Compounds & Radiopharmaceuticals, 29, 867-867 (1991)
Enzyme-catalyzed asymmetric Mannich reaction using acylase from Aspergillus melleus
Journal of Molecular Catalysis. B, Enzymatic, 111, 16-20 (2015)
Physicochemical and catalytic properties of acylase I from aspergillus melleus immobilized on amino- and carbonyl-grafted stober silica
Biotechnology Progress, 767-777 (2018)
Preparation of the enantiomers of 1-phenylethan-1, 2-diol. Regio-and enantioselectivity of acylase I and Candida antarctica lipases A and B
Tetrahedron, 12(17), 2447-2455 (2001)
Antonie van Leeuwenhoek, 94(4), 527-532 (2008-07-31)
A total of 190 Campylobacter spp. isolates, of which 34 gave the result of very weak activity, and 156 gave the negative activity in the test for hippurate hydrolysis were characterized. The genomic DNA was isolated from a fresh culture
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service