Skip to Content
Merck
All Photos(1)

Key Documents

P7127

Sigma-Aldrich

Phospho(enol)pyruvic acid monopotassium salt

≥97% (enzymatic), powder

Synonym(s):

2-(Phosphonooxy)-2-propenoic acid monopotassium salt, mono-Potassium phosphoenolpyruvate, PEP-K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4KO6P
CAS Number:
Molecular Weight:
206.13
Beilstein:
4603446
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Phospho(enol)pyruvic acid monopotassium salt, ≥97% (enzymatic)

Quality Level

Assay

≥97% (enzymatic)

form

powder

solubility

water: 100 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[K+].OC(=O)C(=C)OP(O)([O-])=O

InChI

1S/C3H5O6P.K/c1-2(3(4)5)9-10(6,7)8;/h1H2,(H,4,5)(H2,6,7,8);/q;+1/p-1

InChI key

SOSDSEAIODNVPX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phospho(enol)pyruvic acid (PEP) acts as an inhibitor of hexokinase, phosphoglucose isomerase, phosphofructokinase and aldolase. It is a bifunctional carbohydrate, which exhibits antioxidant property. PEP may be a potential organ preservation agent in clinical transplantation.

Application

Phospho(enol)pyruvic acid monopotassium salt has been used:
  • to maintain a constant concentration of adenosine triphosphate (ATP) for active gel assembly
  • as a component in assay buffer for in vitro ATPase assay
  • as a stock solution for optical trapping assay

Biochem/physiol Actions

Phospho(enol)pyruvic acid (PEP) is involved in glycolysis and gluconeogeneis. In glycolysis, PEP is metabolized by Pyruvate Kinase to yield pyruvate. In plants, PEP is involved in the formation of aromatic amino acids as well as in the carbon fixation pathway.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single Molecule Analysis (2018)
W J Bettger et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(9), 5588-5592 (1981-09-01)
A serum-free, hormone-supplemented medium, enriched with a mixture of lipids, has been developed that supports rapid clonal growth of human diploid fetal lung fibroblasts (Flow 2000, WI-38, MRC-5, and IMR-90) and of low-passage human foreskin fibroblasts. The medium, which contains
Phosphoenolpyruvic acid, an intermediate of glycolysis, attenuates cellular injury induced by hydrogen peroxide and 2-deoxy-D-glucose in the porcine proximal kidney tubular cell line, LLC-PK1
Kondo Y, et al.
Journal of Health Science, 56(6), 727-732 (2010)
Alan Cahill et al.
Methods in molecular biology (Clifton, N.J.), 447, 381-394 (2008-03-29)
Chronic alcohol consumption has been shown to severely compromise mitochondrial protein synthesis. Hepatic mitochondria isolated from alcoholic animals contain decreased levels of respiratory complexes and display depressed respiration rates when compared to pair-fed controls. One underlying mechanism for this involves
Investigation of the reaction between phosphoenolpyruvic acid and thiosulfate-nitrosyl iron complex
Zanina AA, et al.
Russian Chemical Bulletin, 66(5), 927-931 (2017)

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service