Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phospho(enol)pyruvic acid monopotassium salt
Synonym(s):
mono-Potassium phosphoenolpyruvate, PEP monopotassium salt, PEP-K, Phospho(enol)pyruvate potassium salt, 2-(Phosphonooxy)-2-propenoic acid monopotassium salt
Linear Formula:
KOP(O)(OH)OC(=CH
2
)CO
2
H
CAS Number:
4265-07-0
Molecular Weight:
206.13
EC Number:
224-247-0
Products
Product Category
building blocks (1)
substrates (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
860077
Phospho(enol)pyruvic acid monopotassium salt,
99%
Expand
View Pricing
P7127
Phospho(enol)pyruvic acid monopotassium salt,
≥97% (enzymatic), powder
Expand
View Pricing