Skip to Content
Merck
All Photos(3)

Key Documents

P9541

Sigma-Aldrich

Potassium chloride

for molecular biology, ≥99.0%

Synonym(s):

KCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

Assay

≥99.0%

form

powder

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

cation traces

Fe: ≤3 ppm
heavy metals (as Pb): ≤5 ppm

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chloride (KCl) is useful in studies of ion transport and potassium channels. KCl is known to be used in the crystallization of histone core octamers.

Application

Potassium chloride has been used:
  • as a component of Danieau medium and also used in the preparation of phosphate buffer saline
  • in the preparation of electrolyte solution for electrochemical impedance spectroscopy and cyclic voltammetry measurements
  • as a component of HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)-buffered Tyrode′s solution

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reconstitution of nucleosome core particles from recombinant histones and DNA
Test, 375(20), 23-44 (2003)
Fluid osmolarity acutely and differentially modulates lymphatic vessels intrinsic contractions and lymph flow
Solari E, et al.
Frontiers in Physiology, 9(1), 871-871 (2018)
Electrochemical impedance spectroscopic technique with a functionalized microwire sensor for rapid detection of foodbornepathogens
Lu L, et al.
Biosensors And Bioelectronics, 42(1), 492-495 (2013)
Diffusion through channel derivatives of the Escherichia coli FhuA transport protein
Braun M, et al.
European Journal of Biochemistry, 269(20), 4948-4959 (2002)
High-resolution in situ hybridization to whole-mount zebrafish embryos.
Thisse C and Thisse B
Nature Protocols, 3(1), 59-69 (2008)

Protocols

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

TE Buffer; Elution Buffer; 10x Ligation Buffer; 0.5 M PIPES Buffer; Inoue Transformation Buffer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service