Skip to Content
Merck
All Photos(4)

Key Documents

P3029

Sigma-Aldrich

Phencyclidine hydrochloride

Synonym(s):

1-(1-Phenylcyclohexyl)piperidine hydrochloride, PCP hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N · HCl
CAS Number:
Molecular Weight:
279.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

solubility

H2O: 11.2 mg/mL
0.1 M HCl: 18.4 mg/mL
methanol: 30 mg/mL

SMILES string

Cl.C1CCN(CC1)C2(CCCCC2)c3ccccc3

InChI

1S/C17H25N.ClH/c1-4-10-16(11-5-1)17(12-6-2-7-13-17)18-14-8-3-9-15-18;/h1,4-5,10-11H,2-3,6-9,12-15H2;1H

InChI key

BUAJNGPDPGKBGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

σ opioid receptor agonist and psychostimulant; inhibits NMDA glutamate receptor activation by binding to an allosteric site within the ion channel.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Nicotinic) and Glutamate Receptors (Ion Channel Family) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Vesuna et al.
Nature, 586(7827), 87-94 (2020-09-18)
Advanced imaging methods now allow cell-type-specific recording of neural activity across the mammalian brain, potentially enabling the exploration of how brain-wide dynamical patterns give rise to complex behavioural states1-12. Dissociation is an altered behavioural state in which the integrity of
M Ingallinesi et al.
Molecular psychiatry, 20(8), 951-958 (2014-08-27)
Gpr88, an orphan G-protein-coupled receptor, is highly and almost exclusively expressed in the medium spiny projection neurons of the striatum, and may thus participate in the control of motor functions and cognitive processing that are impaired in neuropsychiatric disorders such
Philip L R Gaskin et al.
Psychopharmacology, 231(12), 2533-2545 (2014-01-10)
Schizophrenia is a debilitating disorder comprising positive, negative and cognitive deficits with a poorly defined neurobiological aetiology; therefore, animal models with greater translational reliability are essential to develop improved therapies. This study combines two developmental challenges in rats, neonatal phencyclidine
R H Porter et al.
Journal of neurochemistry, 64(2), 614-623 (1995-02-01)
Quantitative receptor autoradiography was used to examine the regional binding characteristics of a diverse group of N-methyl-D-aspartate (NMDA)-receptor channel blockers that varied in potency 10(5)-fold. Full competition curves were generated in each of six brain regions for 11 different compounds.
Daniel C Javitt
Schizophrenia bulletin, 38(5), 911-913 (2012-09-19)
At present, all medications for schizophrenia function primarily by blocking dopamine D2 receptors. Over 50 years ago, the first observations were made that subsequently led to development of alternative, glutamatergic conceptualizations. This special issue traces the historic development of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service