Skip to Content
Merck
All Photos(1)

Key Documents

O3757

Sigma-Aldrich

Octyl β-D-glucopyranoside solution

≥95% (HPLC), 50 % (w/v) in H2O

Synonym(s):

n-Octyl glucoside, OGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NB.22

description

non-ionic

Quality Level

Assay

≥95% (HPLC)

mol wt

average mol wt 24500-25000

concentration

50 % (w/v) in H2O

aggregation number

84

CMC

20-25

transition temp

cloud point ≥100

application(s)

detection

storage temp.

−20°C

SMILES string

CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

Application

2.5% n-octyl β-D glucopyranoside has been used to homogenize mouse retina
n-Octyl β-D-glucopyranoside (OGP) is a non-ionic detergent which has been used for isoelectric focusing (IEF) and two-dimensional electrophoresis (2D). OGP has been shown to be superior to Triton X-100 for IEF of plant proteins. n-Octyl β-D-glucopyranoside has also been used for concentration of protein from 2D gels for digestion and mass spectroscopy analysis.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Lopez
Journal of chromatography. B, Biomedical sciences and applications, 722(1-2), 191-202 (1999-03-06)
Two-dimensional electrophoresis has rapidly become the method of choice for resolving complex mixtures of proteins. Since the technique was pioneered in 1975, 2-D gel methods have undergone a series of enhancements to optimize resolution and reproducibility. Recent improvements in the
P Dainese Hatt et al.
European journal of biochemistry, 246(2), 336-343 (1997-06-01)
We have developed a gel electrophoresis system that can concentrate proteins from spots cut out of up to 50 two-dimensional electrophoresis gels. During protein concentration, SDS is substituted with a non-ionic detergent (octyl beta-glucopyranoside) which allows digestion and MS analysis
P J Holloway et al.
Analytical biochemistry, 172(1), 8-15 (1988-07-01)
A technique for the analysis of plant proteins from seed, leaf, root, and coleoptile tissues by high resolution two-dimensional electrophoresis is described. This technique is based primarily on the procedure of P. O'Farrell (1975, J. Biol. Chem. 250, 4007-4021); however
Compartment-specific phosphorylation of phosducin in rods underlies adaptation to various levels of illumination.
Song H, et al.
The Journal of Biological Chemistry (2007)
Christine Ménager et al.
Langmuir : the ACS journal of surfaces and colloids, 26(19), 15453-15463 (2010-09-10)
The present study deals with the morphological modifications of giant dioleoyl phosphatidylcholine vesicles (DOPC GUVs) induced by the nonionic surfactant n-octyl β,D-glucopyranoside at sublytic levels, i.e., in the first steps of the vesicle-to-micelle transition process, when surfactant inserts into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service