Skip to Content
Merck
All Photos(1)

Key Documents

P0878

Sigma-Aldrich

O-Phospho-L-serine

≥98% (TLC), powder, glutamate synthase (ferredoxin)] inhibitor

Synonym(s):

L-2-Amino-3-hydroxypropanoic acid 3-phosphate, L-SOP, L-Serine O-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)OCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
185.07
Beilstein:
1726826
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

Product Name

O-Phospho-L-serine,

form

powder

Quality Level

storage condition

under inert gas

mp

190 °C (lit.)

SMILES string

N[C@@H](COP(O)(O)=O)C(O)=O

InChI

1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1

InChI key

BZQFBWGGLXLEPQ-REOHCLBHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

O-Phospho-L-serine has been used:
  • for the expression, purification and phosphorylation of protein
  • to study its chemical shift
  • to spectroscopically determine its pKa values using Raman spectroscopy

Biochem/physiol Actions

Group III metabotropic glutamate receptor agonist (mGluR4, mGluR7, and mGluR8).
O-Phospho-L-serine (L-SOP) is a molecule which resembles phosphatidylserine head group and it mediates partial inhibition of microglial phagocytosis of apoptotic cells. In zebrafish, L-SOP inhibits Müller glia proliferation and blocks cone cell regeneration in the light-damaged retina. In vivo, L-SOP acts as a substrate for CysM, a cysteine synthase from Mycobacterium tuberculosis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Detection of amino acid and peptide phosphate protonation using Raman spectroscopy
Xie Y, et al.
Analytical Biochemistry, 343(2), 223-230 (2005)
Phosphorylation of OPTN by TBK1 enhances its binding to Ub chains and promotes selective autophagy of damaged mitochondria
Richter B, et al.
Proceedings of the National Academy of Sciences of the USA, 113(15), 4039-4044 (2016)
O-phospho-L-serine and the thiocarboxylated sulfur carrier protein CysO-COSH are substrates for CysM, a cysteine synthase from Mycobacterium tuberculosis.
O'Leary SE
Biochemistry, 47(44), 11606-11615 (2008)
The inhibitor of phagocytosis, O-phospho-L-serine, suppresses Muller glia proliferation and cone cell regeneration in the light-damaged zebrafish retina.
Bailey TJ
Experimental Eye Research, 91(5), 601-612 (2010)
Improved peak identification in 31P-NMR spectra of environmental samples with a standardized method and peak library
Cade-Menun BJ
Geoderma, 257, 102-114 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service