Skip to Content
Merck
All Photos(2)

Key Documents

C2196

Sigma-Aldrich

L-Cysteine S-sulfate

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

S-Sulfo-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO5S2
CAS Number:
Molecular Weight:
201.22
Beilstein:
1726832
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Cysteine S-sulfate, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

N[C@@H](CSS(O)(=O)=O)C(O)=O

InChI

1S/C3H7NO5S2/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H,7,8,9)/t2-/m0/s1

InChI key

NOKPBJYHPHHWAN-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Cysteine S-sulfate (LCSS) is used as an NMDA glutamatergic receptor agonist. It serves as a substrate for cystine lyase(s).
Cysteine is one of the functional amino acids that are associated with growth, reproduction, maintenance and immunity. Cysteine is a source of disulfide linkage in protein and is associated with sulfur transport. At physiological pH, cysteine undergoes rapid oxidation to form cystine. Reduced availability of cysteine or cystine, influences leukocyte metabolism. L-Cysteine serves as a precursor for the rate limiting step in glutathione synthesis that occurs in neurons. It donates inorganic sulfate for detoxification reactions. Therefore, L-cysteine might be associated with neuroprotection. It is found to obstruct the entry of heavy metal ions across the blood-brain barrier into the brain. Increased levels of L-cysteine might lead to neurotoxicity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Nakatani et al.
Microbial cell factories, 11, 62-62 (2012-05-23)
Escherichia coli has two L-cysteine biosynthetic pathways; one is synthesized from O-acetyl L-serine (OAS) and sulfate by L-cysteine synthase (CysK), and another is produced via S-sulfocysteine (SSC) from OAS and thiosulfate by SSC synthase (CysM). SSC is converted into L-cysteine
R Nahum-Levy et al.
Biophysical journal, 80(5), 2152-2166 (2001-04-28)
Two distinct forms of desensitization have been characterized for N-methyl-D-aspartate (NMDA) receptors. One form results from a weakening of agonist affinity when channels are activated whereas the other form of desensitization results when channels enter a long-lived nonconducting state. A
Desensitization of NMDA receptor channels is modulated by glutamate agonists.
Nahum-Levy R
Biophysical Journal, 80(5), 2152-2166 (2001)
E C Ramírez et al.
Journal of agricultural and food chemistry, 47(6), 2218-2225 (2000-05-04)
Cystine lyase is the enzyme responsible for off-aroma deterioration in fresh unblanched broccoli. In this research, cystine lyase purification from broccoli has been optimized. Only one protein peak with cystine lyase activity was found during purification. Broccoli cystine lyase was
F Orrego et al.
Neuroscience, 56(3), 539-555 (1993-10-01)
The chemical nature of the central transmitter responsible for fast excitatory events and other related phenomena is analysed against the historical background that has progressively clarified the structure and function of central synapses. One of the problems posed by research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service