Skip to Content
Merck
All Photos(5)

Key Documents

A1910

Sigma-Aldrich

(±)-2-Amino-4-phosphonobutyric acid

solid

Synonym(s):

(±)-AP-4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10NO5P
CAS Number:
Molecular Weight:
183.10
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

form

solid

color

white

SMILES string

NC(CCP(O)(O)=O)C(O)=O

InChI

1S/C4H10NO5P/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)

InChI key

DDOQBQRIEWHWBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(±)-2-Amino-4-phosphonobutyric acid or DL-AP4 is a group III nonselective metabotropic glutamate receptor (mGluR) agonist. L-2-amino-4-phosphonobutyric acid (L-Ap4) displays close to 500-fold selectivity for group III mGluRs.

Application

(±)-2-Amino-4-phosphonobutyric acid may be used as a glutamic acid receptor blocker in perfusion solution in the eye tissue electroretinography studies.

Biochem/physiol Actions

NMDA glutamate receptor antagonist.

Caution

Hygroscopic, store desiccated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H-M Zhang et al.
Neuroscience, 158(2), 875-884 (2008-11-20)
Chronic neuropathic pain remains an unmet clinical problem because it is often resistant to conventional analgesics. Metabotropic glutamate receptors (mGluRs) are involved in nociceptive processing at the spinal level, but their functions in neuropathic pain are not fully known. In
Alex S McKeown et al.
The Journal of physiology, 594(7), 1841-1854 (2015-12-23)
We propose that the end product of chromophore bleaching in rod photoreceptors, all-trans retinol, is part of a feedback loop that increases the sensitivity of the phototransduction cascade in rods. A previously described light-induced hypersensitivity in rods, termed adaptive potentiation
Jared D Chrispell et al.
Investigative ophthalmology & visual science, 59(12), 5116-5124 (2018-10-30)
To define the functional roles of Grk1 and Grk7 in zebrafish cones in vivo. Genome editing was used to generate grk7a and grk1b knockout zebrafish. Electroretinogram (ERG) analyses of the isolated cone mass receptor potential and the b-wave were performed
Richard Fairless et al.
Investigative ophthalmology & visual science, 61(5), 37-37 (2020-05-22)
To determine the influence of RIBEYE deletion and the resulting absence of synaptic ribbons on retinal light signaling by electroretinography. Full-field flash electroretinograms (ERGs) were recorded in RIBEYE knock-out (KO) and wild-type (WT) littermate mice under photopic and scotopic conditions
Satoshi Okado et al.
Investigative ophthalmology & visual science, 58(12), 5518-5525 (2017-10-28)
We determined the effects of a remodeled inner retina on the flicker electroretinograms (ERGs) in a rabbit eye at an advanced stage of inherited retinal degeneration. Six wild-type (WT) and four rhodopsin P347L transgenic (Tg) rabbits were studied at 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service