Skip to Content
Merck
All Photos(1)

Key Documents

P0022

Sigma-Aldrich

PS48

≥98% (HPLC)

Synonym(s):

5-(4-Chloro-phenyl)-3-phenyl-pent-2-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO2
CAS Number:
Molecular Weight:
286.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)\C=C(\CCc1ccc(Cl)cc1)c2ccccc2

InChI

1S/C17H15ClO2/c18-16-10-7-13(8-11-16)6-9-15(12-17(19)20)14-4-2-1-3-5-14/h1-5,7-8,10-12H,6,9H2,(H,19,20)/b15-12-

InChI key

LLJYFDRQFPQGNY-QINSGFPZSA-N

Biochem/physiol Actions

PDK1 (phosphoinositide-dependent protein kinase 1) activator which binds to the HM/PIF binding pocket rather than the ATP-binding site.
PS48 is a PDK1 (phosphoinositide-dependent protein kinase 1) activator which binds to the HM/PIF binding pocket rather than the ATP-binding site. PS48 is one of only a few truly allosteric compounds targeting a regulatory binding site on a protein kinase catalytic domain that is not adjacent to or overlapping with the ATP-binding site.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mito Kanatsu-Shinohara et al.
The Journal of reproduction and development, 64(6), 511-522 (2018-09-04)
Stem cell homing is a complex phenomenon that involves multiple steps; thus far, attempts to increase homing efficiency have met with limited success. Spermatogonial stem cells (SSCs) migrate to the niche after microinjection into seminiferous tubules, but the homing efficiency
Satoshi Watanabe et al.
Biology of reproduction, 100(2), 523-534 (2018-08-31)
Spermatogonial stem cells (SSCs) provide the foundation of spermatogenesis. However, because of their small number and slow self-renewal, transfection of SSCs has met with limited success. Although several viral vectors can infect SSCs, genome integration and an inability to maintain
Steven B Daines et al.
The Journal of arthroplasty, 32(1), 274-279 (2016-08-16)
Increased range of motion to higher degrees of flexion following total knee arthroplasty has been postulated to increase implant damage and revision rates, even in designs modified to accommodate high flexion. We examined posterior-stabilized and high-flexion retrieved tibial inserts to
Risheng Ma et al.
Frontiers in endocrinology, 11, 446-446 (2020-08-08)
Background: The success in rescuing thyroid deficiency in mice using thyroid cells derived from embryonic stem (ES) cells, together with the discovery of human induced pluripotent stem cells (iPSCs) from somatic cells, has raised the possibility of patient-specific thyroid cell
Rui Sun et al.
Cellular and molecular gastroenterology and hepatology, 11(1), 249-272 (2020-08-23)
TNFSF15 genetic variants leading to increased TNF superfamily member 15 (TNFSF15) expression confer risk for inflammatory bowel disease (IBD), and TNFSF15 is being explored as a therapeutic target in IBD patients. Although the focus for TNFSF15-mediated inflammatory outcomes has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service