Skip to Content
Merck
All Photos(1)

Key Documents

P3032

Sigma-Aldrich

Penicillin G sodium salt

powder, suitable for cell culture, BioReagent

Synonym(s):

Benzylpenicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N2NaO4S
CAS Number:
Molecular Weight:
356.37
Beilstein:
3834217
EC Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
34010400
PubChem Substance ID:
NACRES:
NA.76

product name

Penicillin G sodium salt, powder, BioReagent, suitable for cell culture

product line

BioReagent

form

powder

potency

≥1477 units per mg

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 100 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)Cc3ccccc3)C([O-])=O

InChI

1S/C16H18N2O4S.Na/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1

InChI key

FCPVYOBCFFNJFS-LQDWTQKMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam
Solutions should be filter sterilized and stored at 2-8°C for 1 week or at -20°C for extended periods. Solutions are stable at 37°C for 3 days.

Application

Penicillin G is a narrow spectrum natural antibiotic. It is effective against Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.

Biochem/physiol Actions

Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking.

Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.

Caution

Solutions should be filter sterilized and stored at 2-8°C for 1 week or at -20°C for more lengthy periods. Solutions are stable at 37°C for 3 days. The sodium salt is soluble in H2O at 100 mg/mL.

Other Notes

1mu,10mu,25mu,100mu
Keep container tightly closed in a dry and well-ventilated place.Product contains Penicillin Keep in a dry place.

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fleury Augustin Nsole Biteghe et al.
Journal of photochemistry and photobiology. B, Biology, 211, 111982-111982 (2020-09-01)
Aberrant anti-cancer drug efflux mediated by membrane protein ABC transporters (ABCB5 and ABCG2) is thought to characterize melanoma heterogeneous chemoresistant populations, presumed to have unlimited proliferative and self-renewal abilities. Therefore, this study primarily aimed to investigate whether continuous exposure of
Paul P Drury et al.
Neuropharmacology, 83, 62-70 (2014-04-15)
Basal ganglia injury after hypoxia-ischemia remains common in preterm infants, and is closely associated with later cerebral palsy. In the present study we tested the hypothesis that a highly selective neuronal nitric oxide synthase (nNOS) inhibitor, JI-10, would improve survival
Şahin Direkel et al.
Turkiye parazitolojii dergisi, 44(4), 216-220 (2020-12-04)
Leishmaniasis is an important parasitic disease in many countries, including ours. A variety of drugs are currently used for its treatment. However, certain side effects of these drugs, such as teratogenicity, hepatotoxicity and nephrotoxicity, have been reported in some patients.
Elżbieta Łopieńska-Biernat et al.
Pathogens (Basel, Switzerland), 9(12) (2020-12-16)
Anisakis simplex (s.l.) is a complex of three sibling (biological) species of parasitic nematodes of marine mammals, including A. berlandi, A. pegreffii and A. simplex (s.s.). It is characterized by a complex life cycle in which humans can become accidental
Maria Eugenia Sabatino et al.
Frontiers in cell and developmental biology, 7, 297-297 (2019-12-12)
Krüppel-like factor 6 (KLF6) is a transcription factor involved in the regulation of several cellular processes. Regarding its role in tumorigenesis, KLF6 is considered a tumor suppressor. Numerous reports demonstrate its frequent genomic loss or down-regulation, implying a functional inactivation

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service