Skip to Content
Merck
All Photos(4)

Key Documents

L2637

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O55:B5

purified by gel-filtration chromatography

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (O55:B5)

Quality Level

form

lyophilized powder

purified by

gel-filtration chromatography

impurities

<3% Protein

color

white to faint gray

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is extracted from E. coli serotype O55:B5 and purified by gel filtration. The source strain is CDC 1644-70. The LPS O55:B5 has been used to stimulate human peritoneal macrophages at 1 ng/ml and to stimulate equine peritoneal macrophages at 1-100 ng/ml.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Vier et al.
Cell death & disease, 7, e2103-e2103 (2016-02-20)
Neutrophil granulocytes are innate effector cells of the first line of defense against pyogenic bacteria. Neutrophil lifespan is short, is prolonged by pro-inflammatory stimuli, controls functionality of the cells and can determine tissue damage. Experimental analysis of primary neutrophils is
Yunru Ren et al.
Water research, 154, 153-161 (2019-02-21)
Aerosolized reclaimed water can cause inflammatory responses in lung after inhalation, and endotoxin has been identified as the main inducer. Since the effects of disinfection treatments on endotoxins had conflicting results, this study explored the changes of endotoxin activity and
John Bienenstock et al.
PloS one, 8(10), e76236-e76236 (2013-10-08)
Human milk oligosaccharides (HMO) are being studied by different groups exploring a broad range of potential beneficial effects to the breastfed infant. Many of these effects have been attributed to a growth promotion effect on certain gut organisms such as
Zili Zhai et al.
Inflammation, 36(2), 346-354 (2012-10-12)
The nitrone spin trap 5,5-dimethyl-1-pyrroline N-oxide (DMPO) is commonly used to study free radicals. Due to its free radical trapping properties, DMPO is thought to reduce free radial-mediated oxidative damage and other related cellular responses. The purpose of this study
Zili Zhai et al.
Free radical biology & medicine, 53(1), 172-181 (2012-05-15)
The free-radical-operated mechanism of death of activated macrophages at sites of inflammation is unclear, but it is important to define it in order to find targets to prevent further tissue dysfunction. A well-defined model of macrophage activation at sites of

Articles

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Related Content

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service