Skip to Content
Merck
All Photos(2)

Key Documents

N8161

Sigma-Aldrich

NS309

≥98% (HPLC), solid

Synonym(s):

3-Oxime-6,7-dichloro-1H-indole-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4Cl2N2O2
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: 22 mg/mL

storage temp.

2-8°C

SMILES string

O\N=C1\C(=O)Nc2c(Cl)c(Cl)ccc12

InChI

1S/C8H4Cl2N2O2/c9-4-2-1-3-6(5(4)10)11-8(13)7(3)12-14/h1-2,14H,(H,11,12,13)

InChI key

CVOUSAVHMDXCKG-UHFFFAOYSA-N

Biochem/physiol Actions

NS309 is a Ca2+-activated IK/SK potassium channel activator.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jackson T Fontaine et al.
Mechanisms of ageing and development, 192, 111389-111389 (2020-11-01)
Vascular cognitive impairment, the second most common cause of dementia, profoundly affects hippocampal-dependent functions. However, while the growing literature covers complex neuronal interactions, little is known about the sustaining hippocampal microcirculation. Here we examined vasoconstriction to physiological pressures of hippocampal
Fan-Fan Li et al.
Placenta, 58, 9-16 (2017-10-01)
Excessive constriction of placental chorionic plate arteries (CPAs) may be associated with preeclampsia (PE). Nitric oxide (NO) as well as intermediate and small Ca The location of IK IK This study provides evidence that dysregulated IK
Abbie C Johnson et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 37(8), 2857-2869 (2016-11-07)
We investigated the effect of experimental preeclampsia on hyperemia during seizure in the hippocampus and vascular function and structure of hippocampal arterioles using Sprague Dawley rats (n = 14/group) that were nonpregnant, pregnant (d20), or had experimental preeclampsia (induced by a high
Monika Kloza et al.
Vascular pharmacology, 99, 65-73 (2017-10-19)
The aim of this study was to examine the influence of deoxycorticosterone acetate-salt (DOCA-salt) hypertension and chronic treatment with the fatty acid amide hydrolase inhibitor, URB597, on small and intermediate conductance calcium-activated potassium channels and endothelium-dependent hyperpolarization (K
Jensen, B.S., et al.
Abstr. Soc. Neurosci, 2003, 367-367 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service