Skip to Content
Merck
All Photos(1)

Key Documents

S0441

Sigma-Aldrich

SB-366791

≥98% (HPLC), powder

Synonym(s):

N-(3-Methoxyphenyl)-4-chlorocinnamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14NO2Cl
CAS Number:
Molecular Weight:
287.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

originator

GlaxoSmithKline

SMILES string

COc1cccc(NC(=O)\C=C\c2ccc(Cl)cc2)c1

InChI

1S/C16H14ClNO2/c1-20-15-4-2-3-14(11-15)18-16(19)10-7-12-5-8-13(17)9-6-12/h2-11H,1H3,(H,18,19)/b10-7+

InChI key

RYAMDQKWNKKFHD-JXMROGBWSA-N

Gene Information

human ... TRPV1(7442)

Related Categories

Application

SB-366791 has been used as a transient receptor potential cation channel subfamily V member 1 (TRPV1) antagonist:
  • to infer the in vitro and in vivo pharmacology of (E)-3-(4-t-butylphenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide (AMG 9810)
  • to study its effects on sodium hydrogen sulfide (NaHS) or capsaicin-induced contractile activity
  • to study the inhibitory potency of phoneutria toxin (PnTx3-5) (native and recombinant) on various responses mediated by transient receptor potential cation channel subfamily V member 1 (TRPV1)

Biochem/physiol Actions

SB-366791 might exhibit analgesic properties on bone cancer-related pain behavior.
Vanilloid receptor-1 (TRPV1) antagonist.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Niiyama et al.
British journal of anaesthesia, 102(2), 251-258 (2008-11-29)
Bone cancer pain has a major impact on the quality of life of cancer patients but is difficult to treat. Therefore, development of a novel strategy for bone cancer pain is needed for improvement of the patient quality of life.
Riccardo Patacchini et al.
European journal of pharmacology, 509(2-3), 171-177 (2005-03-01)
We have investigated the mechanism through which hydrogen sulfide (H2S) stimulates capsaicin-sensitive primary afferent neurons in the rat isolated urinary bladder. Sodium hydrogen sulfide (NaHS), a donor of H2S, produced concentration-dependent contractile responses (pEC50=3.5+/-0.1) that were unaffected by the transient
Mauricio Guzmán et al.
Immunology, 161(2), 148-161 (2020-07-24)
Dry eye disease (DED) is a highly prevalent ocular surface disorder with neuroimmune pathophysiology. Tear hyperosmolarity (THO), a frequent finding in affected patients, is considered a key element in DED pathogenesis, yet existing animal models are based on subjecting the
SB366791, a TRPV1 antagonist, potentiates analgesic effects of systemic morphine in a murine model of bone cancer pain
Niiyama Y, et al.
British Journal of Anaesthesia, 102(2), 251-258 (2009)
Narender R Gavva et al.
The Journal of pharmacology and experimental therapeutics, 313(1), 474-484 (2004-12-24)
The vanilloid receptor 1 (VR1 or TRPV1) is a membrane-bound, nonselective cation channel expressed by peripheral sensory neurons. TRPV1 antagonists produce antihyperalgesic effects in animal models of inflammatory and neuropathic pain. Here, we describe the in vitro and in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service